Assay Details
| Gene Symbol: SYNPO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166208.1 | NP_001159680.1 | 2 - 3 | 774 | 97 | |
| NM_001166209.1 | NP_001159681.1 | 2 - 3 | 707 | 97 | ||
| XM_006714755.3 | XP_006714818.1 | 2 - 3 | 762 | 97 | ||
| XM_011537552.2 | XP_011535854.1 | 2 - 3 | 759 | 97 | ||
| XM_017009007.1 | XP_016864496.1 | 2 - 3 | 795 | 97 | ||
| XM_017009008.1 | XP_016864497.1 | 2 - 3 | 734 | 97 | ||
| GenBank mRNA | AK127049.1 | - | 2 - 3 | 707 | 97 | |
| AL831818.1 | - | 2 - 3 | 774 | 97 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000833.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000307662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000307662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000307662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000307662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000307662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000307662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000307662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000307662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000394243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000394243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000394243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000394243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000394243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000394243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000394243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000394243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000394243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000394243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000518872.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000518872.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000519664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000519664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000519664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000519664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000519664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000519664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000519664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000519664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000522122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000522122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000522122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000522122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000522122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000522122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000522122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000522122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | ENST00000522122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001109974.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001109974.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001109974.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001109974.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001109974.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001109974.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001109974.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001109974.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001166208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001166208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001166208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001166208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001166208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001166208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001166208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001166208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001166208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001166209.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001166209.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001166209.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001166209.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001166209.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001166209.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001166209.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001166209.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_001166209.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_007286.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_007286.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_007286.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_007286.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_007286.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_007286.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_007286.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009097.hg.1 | NM_007286.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011994.hg.1 | TC05000833.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011994.hg.1 | ENST00000394243.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011994.hg.1 | ENST00000522122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011994.hg.1 | NM_001166208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011994.hg.1 | NM_001166209.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011992.hg.1 | TC05000833.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011992.hg.1 | ENST00000394243.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011992.hg.1 | NM_001166208.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011993.hg.1 | TC05000833.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011993.hg.1 | ENST00000522122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05011993.hg.1 | NM_001166209.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500163571.hg.1 | ENST00000394243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163571.hg.1 | NM_001166208.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500163571.hg.1 | ENST00000394243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163571.hg.1 | NM_001166208.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500163571.hg.1 | TC0500009097.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500163574.hg.1 | ENST00000394243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163574.hg.1 | ENST00000522122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163574.hg.1 | NM_001166208.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500163574.hg.1 | NM_001166209.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500163574.hg.1 | ENST00000394243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163574.hg.1 | ENST00000522122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163574.hg.1 | NM_001166208.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500163574.hg.1 | NM_001166209.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500163574.hg.1 | TC0500009097.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500163572.hg.1 | ENST00000522122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163572.hg.1 | NM_001166209.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500163572.hg.1 | ENST00000522122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163572.hg.1 | NM_001166209.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500163572.hg.1 | TC0500009097.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500163573.hg.1 | ENST00000394243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163573.hg.1 | ENST00000522122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163573.hg.1 | NM_001166208.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500163573.hg.1 | NM_001166209.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500163573.hg.1 | ENST00000394243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163573.hg.1 | ENST00000522122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163573.hg.1 | NM_001166208.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500163573.hg.1 | NM_001166209.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500163573.hg.1 | TC0500009097.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000307662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000307662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000307662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000307662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000307662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000307662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000307662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000307662.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000394243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000394243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000394243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000394243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000394243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000394243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000394243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000394243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000394243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000394243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000518872.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000518872.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000519664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000519664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000519664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000519664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000519664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000519664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000519664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000519664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000522122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000522122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000522122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000522122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000522122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000522122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000522122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000522122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | ENST00000522122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001109974.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001109974.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001109974.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001109974.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001109974.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001109974.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001109974.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001109974.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001166208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001166208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001166208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001166208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001166208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001166208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001166208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001166208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001166208.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001166209.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001166209.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001166209.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001166209.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001166209.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001166209.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001166209.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001166209.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_001166209.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_007286.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_007286.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_007286.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_007286.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_007286.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_007286.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_007286.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009097.hg.1 | NM_007286.5 | Refseq |

SYNPO
Genomic Map