Assay Details
| Gene Symbol: CEACAM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006890.4 | NP_008821.2 | 4 - 5 | 939 | 109 | |
| GenBank mRNA | BC121132.2 | - | 4 - 5 | 833 | 40120888 | 109 |
| X98311.1 | - | 4 - 5 | 829 | 109 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001560.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19021216.hg.1 | ENST00000006724.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021216.hg.1 | NM_006890.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021216.hg.1 | TC19001560.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021217.hg.1 | ENST00000006724.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021217.hg.1 | ENST00000412062.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021217.hg.1 | NM_006890.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021217.hg.1 | TC19001560.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010765.hg.1 | ENST00000006724.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010765.hg.1 | ENST00000006724.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010765.hg.1 | ENST00000006724.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010765.hg.1 | ENST00000006724.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010765.hg.1 | ENST00000006724.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010765.hg.1 | ENST00000006724.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010765.hg.1 | ENST00000006724.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010765.hg.1 | ENST00000006724.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010765.hg.1 | ENST00000006724.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010765.hg.1 | ENST00000006724.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010765.hg.1 | ENST00000401731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010765.hg.1 | ENST00000401731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010765.hg.1 | ENST00000401731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010765.hg.1 | ENST00000401731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010765.hg.1 | ENST00000401731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010765.hg.1 | ENST00000401731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010765.hg.1 | ENST00000401731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010765.hg.1 | ENST00000599715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010765.hg.1 | ENST00000599715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010765.hg.1 | ENST00000599715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010765.hg.1 | ENST00000602225.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010765.hg.1 | ENST00000602225.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010765.hg.1 | ENST00000602225.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010765.hg.1 | NM_001291485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010765.hg.1 | NM_001291485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010765.hg.1 | NM_001291485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010765.hg.1 | NM_001291485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010765.hg.1 | NM_001291485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010765.hg.1 | NM_001291485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010765.hg.1 | NM_001291485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010765.hg.1 | NM_001291485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010765.hg.1 | NM_001291485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021218.hg.1 | ENST00000006724.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021218.hg.1 | ENST00000412062.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021218.hg.1 | NM_006890.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021218.hg.1 | TC19001560.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1900188995.hg.1 | ENST00000006724.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900188995.hg.1 | ENST00000401731.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900188995.hg.1 | NM_001291485.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900188995.hg.1 | TC1900010765.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900188995.hg.1 | ENST00000006724.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900188995.hg.1 | ENST00000401731.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900188995.hg.1 | NM_001291485.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900188997.hg.1 | ENST00000006724.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900188997.hg.1 | ENST00000401731.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900188997.hg.1 | NM_001291485.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900188997.hg.1 | TC1900010765.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900188997.hg.1 | ENST00000006724.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900188997.hg.1 | ENST00000401731.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900188997.hg.1 | NM_001291485.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900188994.hg.1 | ENST00000006724.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900188994.hg.1 | NM_001291485.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900188994.hg.1 | TC1900010765.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900188994.hg.1 | ENST00000006724.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900188994.hg.1 | NM_001291485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010765.hg.1 | ENST00000006724.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010765.hg.1 | ENST00000006724.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010765.hg.1 | ENST00000006724.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010765.hg.1 | ENST00000006724.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010765.hg.1 | ENST00000006724.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010765.hg.1 | ENST00000006724.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010765.hg.1 | ENST00000006724.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010765.hg.1 | ENST00000006724.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010765.hg.1 | ENST00000006724.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010765.hg.1 | ENST00000006724.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010765.hg.1 | ENST00000401731.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010765.hg.1 | ENST00000401731.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010765.hg.1 | ENST00000401731.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010765.hg.1 | ENST00000401731.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010765.hg.1 | ENST00000401731.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010765.hg.1 | ENST00000401731.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010765.hg.1 | ENST00000401731.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010765.hg.1 | ENST00000599715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010765.hg.1 | ENST00000599715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010765.hg.1 | ENST00000599715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010765.hg.1 | ENST00000602225.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010765.hg.1 | ENST00000602225.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010765.hg.1 | ENST00000602225.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010765.hg.1 | NM_001291485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010765.hg.1 | NM_001291485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010765.hg.1 | NM_001291485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010765.hg.1 | NM_001291485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010765.hg.1 | NM_001291485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010765.hg.1 | NM_001291485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010765.hg.1 | NM_001291485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010765.hg.1 | NM_001291485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010765.hg.1 | NM_001291485.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 194623 | - | - |

CEACAM7
Genomic Map