Assay Details
Gene Symbol: OR3A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012373.2 | NP_036505.2 | 1 | 652 | 71 | |
XM_011524028.2 | XP_011522330.1 | 2 | 662 | 71 | ||
GenBank mRNA | BC069415.1 | - | 1 | 652 | 7262255 | 71 |
BC108921.2 | - | 1 | 646 | 40023068 | 71 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10197304 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10197264 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30746759 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30746767 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10197282 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10197292 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10197284 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16936536 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10197311 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10197290 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10197302 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30746786 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10197310 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10197307 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10197271 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30746794 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30746816 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30746779 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30746762 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10197318 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10197299 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30746777 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30746787 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30746778 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30746772 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10197312 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10197303 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30746790 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10197301 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10197272 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30746773 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16936537 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30746820 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10197276 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30746808 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16936523 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10197268 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10197291 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10197274 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30746792 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30746817 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10197319 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10197266 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10197288 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30746763 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10197280 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30746801 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P107483 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P107483 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 34004 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 54936 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P107483 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_23_P107483 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | ERCC-00061_161 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10197271 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10197304 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30746778 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30746772 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10197283 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30746773 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16936537 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30746786 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30746763 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10197280 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30746779 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10197268 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10197307 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10197284 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10197292 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30746808 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10197301 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10197274 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10197276 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10197282 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30746767 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10197303 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16936523 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30746762 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30746759 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30746790 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10197298 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30746793 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10197291 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16936536 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30746794 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30746801 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10197288 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30746792 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30746777 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10197304 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30746779 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30746759 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10197268 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30746792 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10197288 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30746801 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30746773 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30746790 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30746794 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30746778 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30746808 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10197271 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10197274 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16936523 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30746767 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16936536 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10197303 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10197301 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10197298 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10197292 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10197284 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16936537 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30746793 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10197280 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10197283 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10197307 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10197276 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30746772 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30746763 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30746786 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10197291 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30746762 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30746777 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10197282 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_23_P107483 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P10197293 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P10197267 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P10197290 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P10197301 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P10197305 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P10197289 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P10197297 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P10197303 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P10197269 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P10197280 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P10197281 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P10197276 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P10197304 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P10197298 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P10197287 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P10197274 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P10197283 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P10197292 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P10197277 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P10197299 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P10197285 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P10197272 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P107483 | - | - |
Custom Human Agilent Array 11k, Chromosome 6, Expression | 7602 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10197265 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10197277 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10197299 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10197285 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10197281 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16936537 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16936526 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10197307 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10197269 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10197287 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10197304 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10197274 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10197272 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16936523 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10197311 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10197320 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10197297 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10197280 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10197293 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10197292 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10197276 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10197301 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10197303 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10197313 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16936539 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10197315 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10197266 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10197283 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10197317 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_012373_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 8003813 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8003812 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 221426_PM_s_at | - | - |
Affymetrix Human Genome U219 Array | 11735981_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11735981_at | - | - |
Human Genome U133 Plus 2.0 Array | 221426_s_at | - | - |
Human Genome U133 Plus 2.0 Array | 221426_PM_s_at | - | - |
Human Genome U133A 2.0 Array | 221426_s_at | - | - |
Human Genome U133A 2.0 Array | 221426_PM_s_at | - | - |
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_012373_at | - | - |
AffymetrixPathwork Diagnostics Pathchip | pw576323 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 221426_s_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 221426_s_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 221426_s_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 221426_s_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 221426_s_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 221426_s_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 221426_s_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 221426_s_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 221426_s_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 221426_s_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 221426_s_at1 | - | - |
Human Genome U133 Set | 221426_PM_s_at | - | - |
Human Genome U133 Set | 221426_s_at | - | - |
Human Genome U133 Set | 234781_PM_at | - | - |
Human Genome U133 Set | 234781_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A008972 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A008972 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 181287 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1801987 | NM_012373.2 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1801987 | BC069415.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1801987 | BC108921.2 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1801987 | NM_012373.2 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1801987 | BC069415.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1801987 | BC108921.2 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1801987 | NM_012373.2 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1801987 | BC069415.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1801987 | BC108921.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1801987 | NM_012373.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1801987 | BC069415.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1801987 | BC108921.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1801987 | NM_012373.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1801987 | BC069415.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1801987 | BC108921.2 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_13562103-S | BC069415.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_13562103-S | BC108921.2 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_16480-S | - | - |
Illumina human-6 v1.0 expression beadchip | 0004120048 | - | - |
Illumina humanRef-8 v1.0 expression beadchip | 0004120048 | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1801987 | NM_012373.2 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1801987 | BC069415.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1801987 | BC108921.2 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1801987 | NM_012373.2 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1801987 | BC069415.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1801987 | BC108921.2 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | ILMN_16480 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_16480 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1801987 | NM_012373.2 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1801987 | BC069415.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1801987 | BC108921.2 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1801987 | NM_012373.2 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1801987 | BC069415.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1801987 | BC108921.2 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1801987 | NM_012373.2 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1801987 | BC069415.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1801987 | BC108921.2 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1801987 | NM_012373.2 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1801987 | BC069415.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1801987 | BC108921.2 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1801987 | NM_012373.2 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1801987 | BC069415.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1801987 | BC108921.2 | human_transcriptome |
