Assay Details
| Gene Symbol: C3orf84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080528.2 | NP_001073997.2 | 2 - 3 | 225 | 67 | |
| XM_011534015.2 | XP_011532317.1 | 2 - 3 | 1049 | 67 | ||
| XM_011534018.2 | XP_011532320.1 | 2 - 3 | 1050 | 67 | ||
 Back To Top
 Back To TopMore Information
Updated Mapping Information
 This assay targets the same location as Clariom D transcript cluster(s) TC0300011055.hg.1
					            	This assay targets the same location as Clariom D transcript cluster(s) TC0300011055.hg.1This assay targets the same location as HTA transcript cluster(s) TC03001405.hg.1
| Set Membership: |  Amplicon length less than or equal to 70  3' Most  Recommended Assay  Homo sapiens  Probe spans exons | 
MicroArray Probes:
					
						
							Affymetrix
					
					
					
						
							
								
						
					
				
			| Array Name | Probe IDs | Targets | Target Type | 
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300011055.hg.1 | ENST00000432035.2 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0300011055.hg.1 | ENST00000432035.2 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0300011055.hg.1 | ENST00000432035.2 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0300011055.hg.1 | ENST00000432035.2 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0300011055.hg.1 | ENST00000432035.2 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0300011055.hg.1 | ENST00000443990.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0300011055.hg.1 | ENST00000443990.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0300011055.hg.1 | ENST00000443990.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0300011055.hg.1 | ENST00000545770.6 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0300011055.hg.1 | ENST00000545770.6 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0300011055.hg.1 | ENST00000545770.6 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0300011055.hg.1 | ENST00000545770.6 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0300011055.hg.1 | ENST00000545770.6 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0300011055.hg.1 | ENST00000545770.6 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | TC0300011055.hg.1 | NM_001080528.2 | Refseq | 
| Affymetrix Human Transcriptome Array 2.0 | TC0300011055.hg.1 | NM_001080528.2 | Refseq | 
| Affymetrix Human Transcriptome Array 2.0 | TC0300011055.hg.1 | NM_001080528.2 | Refseq | 
| Affymetrix Human Transcriptome Array 2.0 | TC0300011055.hg.1 | NM_001080528.2 | Refseq | 
| Affymetrix Human Transcriptome Array 2.0 | TC0300011055.hg.1 | NM_001080528.2 | Refseq | 
| Affymetrix Human Transcriptome Array 2.0 | TC0300011055.hg.1 | NM_001080528.2 | Refseq | 
| Affymetrix Human Transcriptome Array 2.0 | PSR03024930.hg.1 | TC03001405.hg.1 | HTA TC | 
| Affymetrix Human Transcriptome Array 2.0 | PSR03024930.hg.1 | ENST00000432035.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | PSR03024930.hg.1 | ENST00000545770.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | PSR03024930.hg.1 | NM_001080528.2 | Refseq | 
| Affymetrix Human Transcriptome Array 2.0 | PSR03024928.hg.1 | TC03001405.hg.1 | HTA TC | 
| Affymetrix Human Transcriptome Array 2.0 | PSR03024928.hg.1 | ENST00000432035.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | PSR03024928.hg.1 | ENST00000443990.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | PSR03024928.hg.1 | ENST00000545770.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | PSR03024928.hg.1 | NM_001080528.2 | Refseq | 
| Affymetrix Human Transcriptome Array 2.0 | PSR03024932.hg.1 | TC03001405.hg.1 | HTA TC | 
| Affymetrix Human Transcriptome Array 2.0 | PSR03024932.hg.1 | ENST00000545770.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | PSR03024932.hg.1 | NM_001080528.2 | Refseq | 
| Affymetrix Human Transcriptome Array 2.0 | PSR03024931.hg.1 | TC03001405.hg.1 | HTA TC | 
| Affymetrix Human Transcriptome Array 2.0 | PSR03024931.hg.1 | ENST00000545770.1 | Ensembl | 
| Affymetrix Human Transcriptome Array 2.0 | PSR03024931.hg.1 | NM_001080528.2 | Refseq | 
| Affymetrix Human Clariom D Assay | PSR0300181772.hg.1 | ENST00000545770.6 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0300181772.hg.1 | NM_001080528.2 | Refseq | 
| Affymetrix Human Clariom D Assay | PSR0300181772.hg.1 | ENST00000545770.6 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0300181772.hg.1 | NM_001080528.2 | Refseq | 
| Affymetrix Human Clariom D Assay | PSR0300181772.hg.1 | TC0300011055.hg.1 | Clariom D TC | 
| Affymetrix Human Clariom D Assay | TC0300011055.hg.1 | ENST00000432035.2 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0300011055.hg.1 | ENST00000432035.2 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0300011055.hg.1 | ENST00000432035.2 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0300011055.hg.1 | ENST00000432035.2 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0300011055.hg.1 | ENST00000432035.2 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0300011055.hg.1 | ENST00000443990.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0300011055.hg.1 | ENST00000443990.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0300011055.hg.1 | ENST00000443990.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0300011055.hg.1 | ENST00000545770.6 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0300011055.hg.1 | ENST00000545770.6 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0300011055.hg.1 | ENST00000545770.6 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0300011055.hg.1 | ENST00000545770.6 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0300011055.hg.1 | ENST00000545770.6 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0300011055.hg.1 | ENST00000545770.6 | Ensembl | 
| Affymetrix Human Clariom D Assay | TC0300011055.hg.1 | NM_001080528.2 | Refseq | 
| Affymetrix Human Clariom D Assay | TC0300011055.hg.1 | NM_001080528.2 | Refseq | 
| Affymetrix Human Clariom D Assay | TC0300011055.hg.1 | NM_001080528.2 | Refseq | 
| Affymetrix Human Clariom D Assay | TC0300011055.hg.1 | NM_001080528.2 | Refseq | 
| Affymetrix Human Clariom D Assay | TC0300011055.hg.1 | NM_001080528.2 | Refseq | 
| Affymetrix Human Clariom D Assay | TC0300011055.hg.1 | NM_001080528.2 | Refseq | 
| Affymetrix Human Clariom D Assay | PSR0300181769.hg.1 | ENST00000432035.2 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0300181769.hg.1 | ENST00000545770.6 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0300181769.hg.1 | NM_001080528.2 | Refseq | 
| Affymetrix Human Clariom D Assay | PSR0300181769.hg.1 | ENST00000432035.2 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0300181769.hg.1 | ENST00000545770.6 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0300181769.hg.1 | NM_001080528.2 | Refseq | 
| Affymetrix Human Clariom D Assay | PSR0300181769.hg.1 | TC0300011055.hg.1 | Clariom D TC | 
| Affymetrix Human Clariom D Assay | PSR0300181770.hg.1 | ENST00000432035.2 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0300181770.hg.1 | ENST00000545770.6 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0300181770.hg.1 | NM_001080528.2 | Refseq | 
| Affymetrix Human Clariom D Assay | PSR0300181770.hg.1 | ENST00000432035.2 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0300181770.hg.1 | ENST00000545770.6 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0300181770.hg.1 | NM_001080528.2 | Refseq | 
| Affymetrix Human Clariom D Assay | PSR0300181770.hg.1 | TC0300011055.hg.1 | Clariom D TC | 
| Affymetrix Human Clariom D Assay | PSR0300181767.hg.1 | ENST00000432035.2 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0300181767.hg.1 | ENST00000443990.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0300181767.hg.1 | ENST00000545770.6 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0300181767.hg.1 | NM_001080528.2 | Refseq | 
| Affymetrix Human Clariom D Assay | PSR0300181767.hg.1 | ENST00000432035.2 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0300181767.hg.1 | ENST00000443990.1 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0300181767.hg.1 | ENST00000545770.6 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0300181767.hg.1 | NM_001080528.2 | Refseq | 
| Affymetrix Human Clariom D Assay | PSR0300181767.hg.1 | TC0300011055.hg.1 | Clariom D TC | 
| Affymetrix Human Clariom D Assay | PSR0300181771.hg.1 | ENST00000432035.2 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0300181771.hg.1 | ENST00000545770.6 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0300181771.hg.1 | NM_001080528.2 | Refseq | 
| Affymetrix Human Clariom D Assay | PSR0300181771.hg.1 | ENST00000432035.2 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0300181771.hg.1 | ENST00000545770.6 | Ensembl | 
| Affymetrix Human Clariom D Assay | PSR0300181771.hg.1 | NM_001080528.2 | Refseq | 
| Affymetrix Human Clariom D Assay | PSR0300181771.hg.1 | TC0300011055.hg.1 | Clariom D TC | 
 Back To Top
 Back To Top
 C3orf84
C3orf84 Genomic Map
Genomic Map Back To Top
 Back To Top