Assay Details
| Gene Symbol: C3orf84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080528.2 | NP_001073997.2 | 3 - 4 | 290 | 67 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001405.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02225782 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02225787 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02225789 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22360318 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02225778 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02225785 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02225826 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02225803 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02225841 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02225810 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22360312 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02225824 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02225818 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22360342 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22360339 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02225823 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22360317 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02225790 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02225804 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02225835 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02225825 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22360337 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02225812 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22360316 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02225822 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02225819 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22360296 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02225784 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02225795 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02225802 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22360304 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02225801 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02225799 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02225794 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02225833 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02225837 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02225783 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02225830 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02225829 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02225816 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02225827 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 138484 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 144451 | AK131099.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 144451 | AY356349.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 144451 | NM_183374.2 | refseq_mrna_human_NM |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 55356 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 334958 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 276643 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3243345 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_1490 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_1491 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 14324 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P16217711 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P16217711 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3243345 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3243345 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02225822 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02225837 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02225823 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02225814 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02225830 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02225826 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02225818 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02225827 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02225841 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22360342 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02225812 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02225824 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02225819 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22360337 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02225829 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02225809 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02225816 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02225826 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02225818 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02225814 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02225822 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02225837 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02225819 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02225841 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02225809 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22360337 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22360342 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02225830 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02225827 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02225816 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02225824 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02225829 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02225812 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02225823 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 17711 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 96670 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00700556 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P16217711 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00700551 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00700552 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P30993293 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225797 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15435930 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225726 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225761 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225709 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225754 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225776 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225840 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225732 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225821 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225842 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225814 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225729 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225793 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225850 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225807 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15435923 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225783 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225782 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225723 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225751 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225837 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225749 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225707 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225823 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225756 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225717 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225812 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225715 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225769 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225845 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225826 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225711 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225829 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225706 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225773 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225765 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225847 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225790 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225748 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225801 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225848 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225766 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225735 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15435926 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225816 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225787 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225778 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225746 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225713 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225739 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225743 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225720 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225799 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225724 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225834 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225805 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225704 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225741 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225763 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225785 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225771 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225809 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225818 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02225758 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P00700551 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P16217711 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Genome U219 Array | 11735532_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11735532_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR03024925.hg.1 | TC03001405.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03024925.hg.1 | ENST00000443990.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03024925.hg.1 | ENST00000545770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03024925.hg.1 | NM_001080528.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03024926.hg.1 | TC03001405.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03024926.hg.1 | ENST00000432035.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03024926.hg.1 | ENST00000443990.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03024926.hg.1 | ENST00000545770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03024926.hg.1 | NM_001080528.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011055.hg.1 | ENST00000432035.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011055.hg.1 | ENST00000432035.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011055.hg.1 | ENST00000432035.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011055.hg.1 | ENST00000432035.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011055.hg.1 | ENST00000432035.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011055.hg.1 | ENST00000443990.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011055.hg.1 | ENST00000443990.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011055.hg.1 | ENST00000443990.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011055.hg.1 | ENST00000545770.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011055.hg.1 | ENST00000545770.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011055.hg.1 | ENST00000545770.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011055.hg.1 | ENST00000545770.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011055.hg.1 | ENST00000545770.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011055.hg.1 | ENST00000545770.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011055.hg.1 | NM_001080528.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011055.hg.1 | NM_001080528.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011055.hg.1 | NM_001080528.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011055.hg.1 | NM_001080528.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011055.hg.1 | NM_001080528.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011055.hg.1 | NM_001080528.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011055.hg.1 | ENST00000432035.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011055.hg.1 | ENST00000432035.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011055.hg.1 | ENST00000432035.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011055.hg.1 | ENST00000432035.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011055.hg.1 | ENST00000432035.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011055.hg.1 | ENST00000443990.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011055.hg.1 | ENST00000443990.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011055.hg.1 | ENST00000443990.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011055.hg.1 | ENST00000545770.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011055.hg.1 | ENST00000545770.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011055.hg.1 | ENST00000545770.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011055.hg.1 | ENST00000545770.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011055.hg.1 | ENST00000545770.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011055.hg.1 | ENST00000545770.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011055.hg.1 | NM_001080528.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011055.hg.1 | NM_001080528.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011055.hg.1 | NM_001080528.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011055.hg.1 | NM_001080528.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011055.hg.1 | NM_001080528.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011055.hg.1 | NM_001080528.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300181765.hg.1 | ENST00000432035.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300181765.hg.1 | ENST00000443990.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300181765.hg.1 | ENST00000545770.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300181765.hg.1 | NM_001080528.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300181765.hg.1 | ENST00000432035.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300181765.hg.1 | ENST00000443990.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300181765.hg.1 | ENST00000545770.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300181765.hg.1 | NM_001080528.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300181765.hg.1 | TC0300011055.hg.1 | Clariom D TC |
| Human Genome U95 Set | 51371_at | - | - |
| Human Genome U95 Set | 51371_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A021062 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A021062 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 186380 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1723465 | - | - |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1687170 | - | - |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1723465 | - | - |
| HumanHT-12 V4.0 expression beadchip | ILMN_1723465 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1723465 | - | - |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1723465 | - | - |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1723465 | - | - |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1723465 | - | - |

C3orf84
Genomic Map