Assay Details
Gene Symbol: SSTR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001053.3 | NP_001044.1 | 1 | 213 | 130 | |
NM_001172560.1 | NP_001166031.1 | 2 | 196 | 130 | ||
GenBank mRNA | AK308381.1 | - | 1 | 213 | 130 | |
DB202230.1 | - | 1 | 213 | 130 | ||
DQ448303.1 | - | 1 | 125 | 130 | ||
DQ448304.1 | - | 1 | 125 | 130 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
