Assay Details
| Gene Symbol: MBL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000242.2 | NP_000233.1 | 1 - 2 | 250 | 112 | |
| XM_006717861.3 | XP_006717924.1 | 2 - 3 | 318 | 112 | ||
| XM_011539816.2 | XP_011538118.1 | 2 - 3 | 302 | 112 | ||
| GenBank mRNA | AF360991.1 | - | 1 - 2 | 201 | 112 | |
| AY826184.1 | - | 1 - 2 | 185 | 112 | ||
| BC069338.1 | - | 1 - 2 | 235 | 7262231 | 112 | |
| BC096179.3 | - | 1 - 2 | 216 | 40003735 | 112 | |
| BC096180.3 | - | 1 - 2 | 216 | 40003741 | 112 | |
| BC096181.3 | - | 1 - 2 | 216 | 40003742 | 112 | |
| BC096182.3 | - | 1 - 2 | 216 | 40003743 | 112 | |
| X15422.1 | - | 1 - 2 | 250 | 112 | ||
More Information
Set Membership: |
|

MBL2
Genomic Map