Assay Details
| Gene Symbol: RPL11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000975.3 | NP_000966.2 | 1 - 2 | 51 | 80 | |
| GenBank mRNA | AK311816.1 | - | 1 - 2 | 29 | 80 | |
| BF240826.1 | - | 1 - 2 | 52 | 80 | ||
| BU954396.1 | - | 1 - 2 | 44 | 80 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000299.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK311816.1,NM_000975.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
ribosomal large subunit assembly
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
rRNA processing
translation
translational initiation
protein targeting
SRP-dependent cotranslational protein targeting to membrane
viral transcription
protein localization to nucleus
ribosomal large subunit biogenesis
RNA binding
structural constituent of ribosome
protein binding
rRNA binding
poly(A) RNA binding
ribosomal large subunit assembly
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
rRNA processing
translation
translational initiation
protein targeting
SRP-dependent cotranslational protein targeting to membrane
viral transcription
protein localization to nucleus
ribosomal large subunit biogenesis
RNA binding
structural constituent of ribosome
protein binding
rRNA binding
poly(A) RNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200012847.hg.1 | ENST00000455212.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008598.hg.1 | ENST00000498453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007326.hg.1 | ENST00000374550.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007326.hg.1 | ENST00000374550.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007326.hg.1 | ENST00000374550.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007326.hg.1 | ENST00000374550.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007326.hg.1 | ENST00000374550.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007326.hg.1 | ENST00000374550.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007326.hg.1 | ENST00000374550.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007326.hg.1 | ENST00000443624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007326.hg.1 | ENST00000443624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007326.hg.1 | ENST00000443624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007326.hg.1 | ENST00000443624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007326.hg.1 | ENST00000458455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007326.hg.1 | ENST00000458455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007326.hg.1 | ENST00000458455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007326.hg.1 | ENST00000458455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007326.hg.1 | ENST00000458455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007326.hg.1 | ENST00000467075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007326.hg.1 | ENST00000467075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007326.hg.1 | ENST00000482370.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007326.hg.1 | ENST00000482370.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007326.hg.1 | ENST00000482370.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007326.hg.1 | ENST00000482370.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007326.hg.1 | ENST00000482370.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007326.hg.1 | ENST00000482370.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007326.hg.1 | ENST00000482370.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007326.hg.1 | NM_000975.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007326.hg.1 | NM_000975.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007326.hg.1 | NM_000975.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007326.hg.1 | NM_000975.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007326.hg.1 | NM_000975.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007326.hg.1 | NM_000975.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007326.hg.1 | NM_000975.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007326.hg.1 | NM_001199802.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007326.hg.1 | NM_001199802.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007326.hg.1 | NM_001199802.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007326.hg.1 | NM_001199802.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007326.hg.1 | NM_001199802.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007326.hg.1 | NM_001199802.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007326.hg.1 | NM_001199802.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009678.hg.1 | ENST00000603489.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008366.hg.1 | ENST00000395256.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011429.hg.1 | ENST00000332269.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01004533.hg.1 | TC01000299.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01004533.hg.1 | ENST00000374550.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01004533.hg.1 | NM_000975.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01004533.hg.1 | NM_001199802.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012173.hg.1 | ENST00000495697.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151008.hg.1 | ENST00000374550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151008.hg.1 | ENST00000443624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151008.hg.1 | ENST00000458455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151008.hg.1 | ENST00000467075.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151008.hg.1 | ENST00000482370.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151008.hg.1 | NM_000975.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100151008.hg.1 | NM_001199802.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100151008.hg.1 | ENST00000374550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151008.hg.1 | ENST00000443624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151008.hg.1 | ENST00000458455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151008.hg.1 | ENST00000467075.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151008.hg.1 | ENST00000482370.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151008.hg.1 | NM_000975.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100151008.hg.1 | NM_001199802.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100151008.hg.1 | TC0100007326.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100007326.hg.1 | ENST00000374550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007326.hg.1 | ENST00000374550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007326.hg.1 | ENST00000374550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007326.hg.1 | ENST00000374550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007326.hg.1 | ENST00000374550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007326.hg.1 | ENST00000374550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007326.hg.1 | ENST00000374550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007326.hg.1 | ENST00000443624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007326.hg.1 | ENST00000443624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007326.hg.1 | ENST00000443624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007326.hg.1 | ENST00000443624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007326.hg.1 | ENST00000458455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007326.hg.1 | ENST00000458455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007326.hg.1 | ENST00000458455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007326.hg.1 | ENST00000458455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007326.hg.1 | ENST00000458455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007326.hg.1 | ENST00000467075.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007326.hg.1 | ENST00000467075.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007326.hg.1 | ENST00000482370.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007326.hg.1 | ENST00000482370.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007326.hg.1 | ENST00000482370.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007326.hg.1 | ENST00000482370.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007326.hg.1 | ENST00000482370.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007326.hg.1 | ENST00000482370.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007326.hg.1 | ENST00000482370.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007326.hg.1 | NM_000975.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007326.hg.1 | NM_000975.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007326.hg.1 | NM_000975.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007326.hg.1 | NM_000975.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007326.hg.1 | NM_000975.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007326.hg.1 | NM_000975.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007326.hg.1 | NM_000975.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007326.hg.1 | NM_001199802.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007326.hg.1 | NM_001199802.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007326.hg.1 | NM_001199802.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007326.hg.1 | NM_001199802.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007326.hg.1 | NM_001199802.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007326.hg.1 | NM_001199802.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007326.hg.1 | NM_001199802.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011429.hg.1 | ENST00000332269.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151001.hg.1 | ENST00000374550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151001.hg.1 | NM_000975.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100151001.hg.1 | NM_001199802.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100151001.hg.1 | ENST00000374550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151001.hg.1 | NM_000975.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100151001.hg.1 | NM_001199802.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100151001.hg.1 | TC0100007326.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200012847.hg.1 | ENST00000455212.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009678.hg.1 | ENST00000603489.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008598.hg.1 | ENST00000498453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008366.hg.1 | ENST00000395256.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012173.hg.1 | ENST00000495697.1 | Ensembl |

RPL11
Genomic Map