Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017819.hg.1 |
TC09001429.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017819.hg.1 |
ENST00000277216.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017819.hg.1 |
ENST00000545903.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017819.hg.1 |
NM_001001919.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011064.hg.1 |
ENST00000277216.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011064.hg.1 |
NM_001001919.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900175408.hg.1 |
ENST00000277216.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900175408.hg.1 |
NM_001001919.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900175408.hg.1 |
ENST00000277216.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900175408.hg.1 |
NM_001001919.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900175408.hg.1 |
TC0900011064.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900011064.hg.1 |
ENST00000277216.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011064.hg.1 |
NM_001001919.1 |
Refseq |