Assay Details
| Gene Symbol: LRIF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006945.1 | NP_001006946.1 | 2 - 3 | 599 | 89 | |
| NM_018372.3 | NP_060842.3 | 3 - 4 | 2127 | 89 | ||
| XM_011541768.1 | XP_011540070.1 | 3 - 4 | 2127 | 89 | ||
| GenBank mRNA | AK001826.1 | - | 2 - 3 | 705 | 89 | |
| AK002131.1 | - | 2 - 3 | 1068 | 89 | ||
| AL834466.1 | - | 2 - 3 | 1045 | 89 | ||
| AY190122.1 | - | 3 - 4 | 2117 | 89 | ||
| BC008115.1 | - | 2 - 3 | 1269 | 2900378 | 89 | |
| BU664417.1 | - | 2 - 3 | 588 | 89 | ||
More Information
Set Membership: |
|

LRIF1
Genomic Map