Assay Details
Gene Symbol: RPS17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001021.4 | NP_001012.1 | 1 - 2 | 123 | 98 | |
NR_111944.1 | 1 - 2 | 123 | 98 | |||
GenBank mRNA | BC022370.1 | - | 1 - 2 | 36 | 4738331 | 98 |
BC062715.1 | - | 1 - 2 | 121 | 4068842 | 98 | |
BM017772.1 | - | 1 - 2 | 124 | 98 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC15001743.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC022370.1,BC062715.1,NM_001021.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
ribosomal small subunit assembly
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
rRNA processing
translation
translational initiation
SRP-dependent cotranslational protein targeting to membrane
viral transcription
erythrocyte homeostasis
ribosomal small subunit biogenesis
structural constituent of ribosome
poly(A) RNA binding
ribosomal small subunit assembly
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
rRNA processing
translation
translational initiation
SRP-dependent cotranslational protein targeting to membrane
viral transcription
erythrocyte homeostasis
ribosomal small subunit biogenesis
structural constituent of ribosome
poly(A) RNA binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR15017058.hg.1 | ENST00000330339.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15017058.hg.1 | ENST00000559187.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15017058.hg.1 | NM_001021.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15017058.hg.1 | TC15001743.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR15017059.hg.1 | ENST00000330339.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15017059.hg.1 | ENST00000559187.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15017059.hg.1 | NM_001021.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15017059.hg.1 | TC15001743.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0600008384.hg.1 | ENST00000405686.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008384.hg.1 | ENST00000405686.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000330244.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000330244.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000330244.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000330244.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000330244.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000330244.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000558397.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000558397.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000558397.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000558397.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000558397.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000558397.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000559273.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000559273.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000559776.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000559776.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000559776.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000559776.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000560229.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000560229.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000560229.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000560229.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000560229.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000560229.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000560612.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000560612.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000560612.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000560612.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000560612.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000560612.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000560612.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000560612.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000560639.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000560639.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000560639.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000560639.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000560639.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000560639.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000561068.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000561068.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000561068.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000561068.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000561068.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000561068.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000561157.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000561157.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000561157.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000561157.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000561157.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000561157.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000561157.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000561157.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000561440.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000561440.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000561440.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000561440.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000561440.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000561440.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000561440.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000561440.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000561440.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010910.hg.1 | ENST00000561440.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016297.hg.1 | ENST00000450126.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011459.hg.1 | ENST00000382740.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011459.hg.1 | ENST00000382740.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012746.hg.1 | ENST00000464337.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012280.hg.1 | ENST00000475609.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008805.hg.1 | ENST00000509298.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012021.hg.1 | ENST00000498293.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012692.hg.1 | ENST00000477300.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011309.hg.1 | ENST00000396783.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007468.hg.1 | ENST00000490847.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007119.hg.1 | ENST00000423610.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15017049.hg.1 | ENST00000330339.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15017049.hg.1 | ENST00000559187.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15017049.hg.1 | NM_001021.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15017049.hg.1 | TC15001743.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR15017050.hg.1 | ENST00000330339.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15017050.hg.1 | ENST00000559187.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15017050.hg.1 | NM_001021.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15017050.hg.1 | TC15001743.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0500008409.hg.1 | ENST00000461663.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011459.hg.1 | ENST00000382740.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011459.hg.1 | ENST00000382740.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008805.hg.1 | ENST00000509298.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008409.hg.1 | ENST00000461663.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007468.hg.1 | ENST00000490847.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500185759.hg.1 | ENST00000330244.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500185759.hg.1 | ENST00000330244.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500185759.hg.1 | TC1500010910.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1100012280.hg.1 | ENST00000475609.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500185743.hg.1 | ENST00000330244.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500185743.hg.1 | ENST00000558397.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500185743.hg.1 | ENST00000559273.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500185743.hg.1 | ENST00000559776.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500185743.hg.1 | ENST00000560229.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500185743.hg.1 | ENST00000560612.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500185743.hg.1 | ENST00000560639.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500185743.hg.1 | ENST00000561068.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500185743.hg.1 | ENST00000561157.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500185743.hg.1 | ENST00000561440.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500185743.hg.1 | ENST00000330244.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500185743.hg.1 | ENST00000558397.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500185743.hg.1 | ENST00000559273.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500185743.hg.1 | ENST00000559776.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500185743.hg.1 | ENST00000560229.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500185743.hg.1 | ENST00000560612.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500185743.hg.1 | ENST00000560639.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500185743.hg.1 | ENST00000561068.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500185743.hg.1 | ENST00000561157.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500185743.hg.1 | ENST00000561440.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500185743.hg.1 | TC1500010910.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100016297.hg.1 | ENST00000450126.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011309.hg.1 | ENST00000396783.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012746.hg.1 | ENST00000464337.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012692.hg.1 | ENST00000477300.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000330244.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000330244.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000330244.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000330244.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000330244.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000330244.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000558397.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000558397.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000558397.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000558397.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000558397.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000558397.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000559273.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000559273.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000559776.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000559776.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000559776.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000559776.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000560229.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000560229.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000560229.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000560229.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000560229.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000560229.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000560612.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000560612.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000560612.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000560612.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000560612.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000560612.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000560612.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000560612.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000560639.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000560639.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000560639.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000560639.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000560639.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000560639.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000561068.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000561068.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000561068.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000561068.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000561068.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000561068.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000561157.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000561157.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000561157.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000561157.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000561157.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000561157.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000561157.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000561157.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000561440.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000561440.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000561440.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000561440.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000561440.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000561440.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000561440.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000561440.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000561440.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010910.hg.1 | ENST00000561440.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007119.hg.1 | ENST00000423610.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008384.hg.1 | ENST00000405686.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008384.hg.1 | ENST00000405686.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012021.hg.1 | ENST00000498293.1 | Ensembl |
