Assay Details
Gene Symbol: RPS17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001021.4 | NP_001012.1 | 3 - 4 | 370 | 101 | |
NR_111943.1 | 2 - 3 | 576 | 101 | |||
NR_111944.1 | 3 - 4 | 370 | 101 | |||
GenBank mRNA | AK311951.1 | - | 3 - 4 | 278 | 101 | |
BC009407.2 | - | 1 | 261 | 4334552 | 101 | |
BC019899.1 | - | 1 | 266 | 3882924 | 101 | |
BC020453.1 | - | 1 | 265 | 101 | ||
BC022370.1 | - | 3 - 4 | 283 | 4738331 | 101 | |
BC049824.1 | - | 1 | 258 | 6212543 | 101 | |
BC062715.1 | - | 3 - 4 | 368 | 4068842 | 101 | |
BC070222.1 | - | 1 | 279 | 6738383 | 101 | |
BC071928.1 | - | 1 | 269 | 5090053 | 101 | |
BE905890.1 | - | 2 - 3 | 246 | 101 | ||
BI597659.1 | - | 2 - 3 | 576 | 101 | ||
BM017772.1 | - | 3 - 4 | 372 | 102 | ||
BM457623.1 | - | 2 - 3 | 263 | 101 | ||
M13932.1 | - | 3 - 4 | 279 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
