| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16017953.hg.1 |
ENST00000326043.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16017953.hg.1 |
ENST00000393350.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16017953.hg.1 |
NM_001031804.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16017953.hg.1 |
NM_005360.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16017953.hg.1 |
TC16001291.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16017952.hg.1 |
ENST00000393350.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16017952.hg.1 |
NM_001031804.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16017952.hg.1 |
TC16001291.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16017956.hg.1 |
ENST00000326043.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16017956.hg.1 |
ENST00000393350.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16017956.hg.1 |
NM_001031804.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16017956.hg.1 |
NM_005360.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16017956.hg.1 |
TC16001291.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16017950.hg.1 |
ENST00000393350.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16017950.hg.1 |
NM_001031804.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16017950.hg.1 |
TC16001291.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16017946.hg.1 |
ENST00000393350.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16017946.hg.1 |
NM_001031804.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16017946.hg.1 |
TC16001291.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16017948.hg.1 |
ENST00000393350.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16017948.hg.1 |
NM_001031804.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16017948.hg.1 |
TC16001291.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16017955.hg.1 |
ENST00000326043.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16017955.hg.1 |
ENST00000393350.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16017955.hg.1 |
NM_001031804.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16017955.hg.1 |
NM_005360.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16017955.hg.1 |
TC16001291.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010951.hg.1 |
ENST00000326043.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010951.hg.1 |
ENST00000326043.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010951.hg.1 |
ENST00000326043.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010951.hg.1 |
ENST00000326043.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010951.hg.1 |
ENST00000326043.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010951.hg.1 |
ENST00000326043.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010951.hg.1 |
ENST00000393350.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010951.hg.1 |
ENST00000393350.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010951.hg.1 |
ENST00000393350.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010951.hg.1 |
ENST00000393350.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010951.hg.1 |
ENST00000393350.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010951.hg.1 |
ENST00000393350.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010951.hg.1 |
ENST00000569649.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010951.hg.1 |
ENST00000569649.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010951.hg.1 |
NM_001031804.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010951.hg.1 |
NM_001031804.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010951.hg.1 |
NM_001031804.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010951.hg.1 |
NM_001031804.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010951.hg.1 |
NM_001031804.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010951.hg.1 |
NM_001031804.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010951.hg.1 |
NM_001031804.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010951.hg.1 |
NM_005360.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010951.hg.1 |
NM_005360.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010951.hg.1 |
NM_005360.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010951.hg.1 |
NM_005360.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010951.hg.1 |
NM_005360.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010951.hg.1 |
NM_005360.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600185131.hg.1 |
ENST00000326043.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600185131.hg.1 |
ENST00000393350.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600185131.hg.1 |
NM_001031804.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600185131.hg.1 |
NM_005360.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600185131.hg.1 |
ENST00000326043.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600185131.hg.1 |
ENST00000393350.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600185131.hg.1 |
NM_001031804.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600185131.hg.1 |
NM_005360.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600185131.hg.1 |
TC1600010951.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1600010951.hg.1 |
ENST00000326043.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010951.hg.1 |
ENST00000326043.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010951.hg.1 |
ENST00000326043.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010951.hg.1 |
ENST00000326043.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010951.hg.1 |
ENST00000326043.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010951.hg.1 |
ENST00000326043.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010951.hg.1 |
ENST00000393350.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010951.hg.1 |
ENST00000393350.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010951.hg.1 |
ENST00000393350.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010951.hg.1 |
ENST00000393350.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010951.hg.1 |
ENST00000393350.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010951.hg.1 |
ENST00000393350.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010951.hg.1 |
ENST00000569649.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010951.hg.1 |
ENST00000569649.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010951.hg.1 |
NM_001031804.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010951.hg.1 |
NM_001031804.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010951.hg.1 |
NM_001031804.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010951.hg.1 |
NM_001031804.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010951.hg.1 |
NM_001031804.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010951.hg.1 |
NM_001031804.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010951.hg.1 |
NM_001031804.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010951.hg.1 |
NM_005360.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010951.hg.1 |
NM_005360.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010951.hg.1 |
NM_005360.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010951.hg.1 |
NM_005360.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010951.hg.1 |
NM_005360.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600010951.hg.1 |
NM_005360.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600185130.hg.1 |
ENST00000326043.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600185130.hg.1 |
ENST00000393350.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600185130.hg.1 |
NM_001031804.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600185130.hg.1 |
NM_005360.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600185130.hg.1 |
ENST00000326043.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600185130.hg.1 |
ENST00000393350.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600185130.hg.1 |
NM_001031804.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600185130.hg.1 |
NM_005360.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600185130.hg.1 |
TC1600010951.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600185127.hg.1 |
ENST00000393350.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600185127.hg.1 |
NM_001031804.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600185127.hg.1 |
ENST00000393350.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600185127.hg.1 |
NM_001031804.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600185127.hg.1 |
TC1600010951.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600185129.hg.1 |
ENST00000326043.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600185129.hg.1 |
ENST00000393350.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600185129.hg.1 |
ENST00000569649.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600185129.hg.1 |
NM_001031804.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600185129.hg.1 |
NM_005360.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600185129.hg.1 |
ENST00000326043.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600185129.hg.1 |
ENST00000393350.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600185129.hg.1 |
ENST00000569649.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600185129.hg.1 |
NM_001031804.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600185129.hg.1 |
NM_005360.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600185129.hg.1 |
TC1600010951.hg.1 |
Clariom D TC |