Assay Details
Gene Symbol: AIPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033054.2 | NP_001028226.1 | 1 - 2 | 196 | 72 | |
NM_001285401.2 | NP_001272330.1 | 1 - 2 | 196 | 72 | ||
NM_001285403.2 | NP_001272332.1 | 1 - 2 | 196 | 72 | ||
NM_014336.4 | NP_055151.3 | 1 - 2 | 196 | 72 | ||
GenBank mRNA | AB593042.1 | - | 1 - 2 | 117 | 72 | |
AB593043.1 | - | 1 - 2 | 117 | 72 | ||
AB593044.1 | - | 1 - 2 | 131 | 72 | ||
AB593045.1 | - | 1 - 2 | 131 | 72 | ||
AB593046.1 | - | 1 - 2 | 133 | 72 | ||
AB593049.1 | - | 1 - 2 | 141 | 72 | ||
AB593050.1 | - | 1 - 2 | 117 | 72 | ||
AB593051.1 | - | 1 - 2 | 118 | 72 | ||
AB593052.1 | - | 1 - 2 | 117 | 72 | ||
AB593053.1 | - | 1 - 2 | 162 | 72 | ||
AB593054.1 | - | 1 - 2 | 105 | 72 | ||
AB593055.1 | - | 1 - 2 | 162 | 72 | ||
AF038437.1 | - | 1 - 2 | 115 | 72 | ||
AF148864.1 | - | 1 - 2 | 72 | |||
AF525970.1 | - | 1 - 2 | 196 | 72 | ||
AJ633678.1 | - | 1 - 2 | 149 | 72 | ||
AJ830743.1 | - | 1 - 2 | 150 | 72 | ||
AK314558.1 | - | 1 - 2 | 117 | 72 | ||
BC012055.1 | - | 1 - 2 | 109 | 4634156 | 72 | |
BX647279.1 | - | 1 - 2 | 162 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
