Assay Details
Gene Symbol: DHRS4L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006720246.3 | XP_006720309.1 | 3 - 4 | 395 | 53 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | ENST00000397065.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | ENST00000397065.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | ENST00000397065.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | ENST00000397065.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | ENST00000397065.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | ENST00000397065.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | ENST00000397065.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | ENST00000397065.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | ENST00000397065.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | ENST00000397065.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | ENST00000558293.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | ENST00000558293.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | ENST00000558293.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | ENST00000558293.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | ENST00000558293.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | ENST00000558293.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | ENST00000558293.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | ENST00000558293.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | ENST00000558293.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | ENST00000558293.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | ENST00000558622.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | ENST00000558622.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | ENST00000558622.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | ENST00000558622.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | ENST00000558622.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | ENST00000558682.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | ENST00000558682.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | ENST00000558682.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | ENST00000558682.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | ENST00000559270.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | ENST00000559270.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | ENST00000559270.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | ENST00000559270.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | ENST00000559270.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | ENST00000559270.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | ENST00000559270.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | ENST00000559270.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NM_001277864.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NM_001277864.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NM_001277864.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NM_001277864.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NM_001277864.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NM_001277864.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NM_001277864.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NM_001277864.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NM_001277864.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102687.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102687.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102687.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102687.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102687.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102687.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102687.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102688.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102688.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102688.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102688.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102688.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102688.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102688.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102688.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102689.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102689.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102689.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102689.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102689.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102689.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102689.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102689.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102689.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102691.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102691.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102691.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102691.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102691.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102691.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102691.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102691.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102692.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102692.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102692.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102692.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102692.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102692.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102692.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102692.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102692.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102692.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102693.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102693.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102693.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102693.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102693.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102693.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102693.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102693.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102693.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102693.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102693.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102693.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006711.hg.1 | NR_102694.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | ENST00000397065.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | ENST00000397065.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | ENST00000397065.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | ENST00000397065.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | ENST00000397065.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | ENST00000397065.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | ENST00000397065.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | ENST00000397065.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | ENST00000397065.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | ENST00000397065.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | ENST00000558293.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | ENST00000558293.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | ENST00000558293.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | ENST00000558293.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | ENST00000558293.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | ENST00000558293.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | ENST00000558293.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | ENST00000558293.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | ENST00000558293.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | ENST00000558293.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | ENST00000558622.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | ENST00000558622.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | ENST00000558622.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | ENST00000558622.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | ENST00000558622.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | ENST00000558682.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | ENST00000558682.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | ENST00000558682.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | ENST00000558682.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | ENST00000559270.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | ENST00000559270.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | ENST00000559270.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | ENST00000559270.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | ENST00000559270.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | ENST00000559270.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | ENST00000559270.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | ENST00000559270.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NM_001277864.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NM_001277864.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NM_001277864.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NM_001277864.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NM_001277864.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NM_001277864.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NM_001277864.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NM_001277864.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NM_001277864.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102687.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102687.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102687.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102687.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102687.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102687.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102687.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102688.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102688.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102688.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102688.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102688.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102688.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102688.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102688.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102689.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102689.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102689.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102689.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102689.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102689.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102689.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102689.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102689.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102691.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102691.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102691.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102691.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102691.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102691.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102691.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102691.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102692.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102692.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102692.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102692.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102692.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102692.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102692.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102692.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102692.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102692.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102693.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102693.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102693.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102693.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102693.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102693.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102693.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102693.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102693.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102693.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102693.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102693.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102694.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102694.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102694.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102694.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102694.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102694.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102694.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102694.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006711.hg.1 | NR_102694.1 | Refseq |
