| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016862.hg.1 |
TC09001346.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016862.hg.1 |
ENST00000375419.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016862.hg.1 |
NM_001098808.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016860.hg.1 |
TC09001346.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016860.hg.1 |
ENST00000375419.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016860.hg.1 |
NM_001098808.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010833.hg.1 |
ENST00000375419.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010833.hg.1 |
ENST00000375419.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010833.hg.1 |
ENST00000375419.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010833.hg.1 |
ENST00000375419.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010833.hg.1 |
ENST00000375419.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010833.hg.1 |
ENST00000375419.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010833.hg.1 |
NM_001098808.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010833.hg.1 |
NM_001098808.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010833.hg.1 |
NM_001098808.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010833.hg.1 |
NM_001098808.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010833.hg.1 |
NM_001098808.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010833.hg.1 |
NM_001098808.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016861.hg.1 |
TC09001346.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016861.hg.1 |
ENST00000375419.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09016861.hg.1 |
NM_001098808.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173709.hg.1 |
ENST00000375419.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900173709.hg.1 |
NM_001098808.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173709.hg.1 |
ENST00000375419.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900173709.hg.1 |
NM_001098808.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173709.hg.1 |
TC0900010833.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900173710.hg.1 |
ENST00000375419.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900173710.hg.1 |
NM_001098808.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173710.hg.1 |
ENST00000375419.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900173710.hg.1 |
NM_001098808.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173710.hg.1 |
TC0900010833.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0900010833.hg.1 |
ENST00000375419.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010833.hg.1 |
ENST00000375419.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010833.hg.1 |
ENST00000375419.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010833.hg.1 |
ENST00000375419.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010833.hg.1 |
ENST00000375419.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010833.hg.1 |
ENST00000375419.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010833.hg.1 |
NM_001098808.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010833.hg.1 |
NM_001098808.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010833.hg.1 |
NM_001098808.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010833.hg.1 |
NM_001098808.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010833.hg.1 |
NM_001098808.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010833.hg.1 |
NM_001098808.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173713.hg.1 |
ENST00000375419.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900173713.hg.1 |
NM_001098808.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173713.hg.1 |
ENST00000375419.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900173713.hg.1 |
NM_001098808.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900173713.hg.1 |
TC0900010833.hg.1 |
Clariom D TC |