Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19008022.hg.1 |
TC19002635.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19008022.hg.1 |
ENST00000356385.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19008022.hg.1 |
ENST00000392002.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19008022.hg.1 |
ENST00000413014.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19008022.hg.1 |
ENST00000436170.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19008022.hg.1 |
ENST00000447302.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19008022.hg.1 |
ENST00000539627.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19008022.hg.1 |
ENST00000542945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19008022.hg.1 |
ENST00000544232.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19008022.hg.1 |
NM_001042595.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19008022.hg.1 |
NM_001098821.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19008022.hg.1 |
NM_001098822.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19008022.hg.1 |
NM_001098823.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19008022.hg.1 |
NM_001098824.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19008022.hg.1 |
NM_001098825.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900201177.hg.1 |
ENST00000356385.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900201177.hg.1 |
ENST00000392002.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900201177.hg.1 |
ENST00000413014.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900201177.hg.1 |
ENST00000436170.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900201177.hg.1 |
ENST00000447302.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900201177.hg.1 |
ENST00000535712.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900201177.hg.1 |
ENST00000539627.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900201177.hg.1 |
ENST00000542945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900201177.hg.1 |
ENST00000544232.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900201177.hg.1 |
ENST00000546050.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900201177.hg.1 |
ENST00000546362.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900201177.hg.1 |
ENST00000604123.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900201177.hg.1 |
NM_001042595.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900201177.hg.1 |
NM_001098821.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900201177.hg.1 |
NM_001098822.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900201177.hg.1 |
NM_001098823.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900201177.hg.1 |
NM_001098824.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900201177.hg.1 |
NM_001098825.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900201177.hg.1 |
TC1900011743.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900201177.hg.1 |
ENST00000356385.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900201177.hg.1 |
ENST00000392002.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900201177.hg.1 |
ENST00000413014.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900201177.hg.1 |
ENST00000436170.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900201177.hg.1 |
ENST00000447302.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900201177.hg.1 |
ENST00000535712.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900201177.hg.1 |
ENST00000539627.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900201177.hg.1 |
ENST00000542945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900201177.hg.1 |
ENST00000544232.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900201177.hg.1 |
ENST00000546050.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900201177.hg.1 |
ENST00000546362.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900201177.hg.1 |
ENST00000604123.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900201177.hg.1 |
NM_001042595.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900201177.hg.1 |
NM_001098821.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900201177.hg.1 |
NM_001098822.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900201177.hg.1 |
NM_001098823.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900201177.hg.1 |
NM_001098824.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900201177.hg.1 |
NM_001098825.1 |
Refseq |