Assay Details
| Gene Symbol: C20orf197 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001302813.1 | NP_001289742.1 | 2 - 3 | 292 | 94 | |
| NM_173644.2 | NP_775915.1 | 3 - 4 | 372 | 94 | ||
| XM_017027828.1 | XP_016883317.1 | 2 - 3 | 9097 | 94 | ||
| XM_017027829.1 | XP_016883318.1 | 3 - 4 | 6759 | 94 | ||
| XM_017027830.1 | XP_016883319.1 | 2 - 3 | 6624 | 94 | ||
| XM_017027831.1 | XP_016883320.1 | 3 - 4 | 8443 | 94 | ||
| XM_017027832.1 | XP_016883321.1 | 3 - 4 | 496 | 94 | ||
| GenBank mRNA | AK091179.1 | - | 3 - 4 | 372 | 94 | |
| BC101349.1 | - | 2 - 3 | 238 | 40025393 | 94 | |
| BC101351.2 | - | 2 - 3 | 238 | 40025395 | 94 | |
| CR744567.1 | - | 2 - 3 | 290 | 94 | ||
More Information
Set Membership: |
|

C20orf197
Genomic Map