Assay Details
Gene Symbol: BEND4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159547.1 | NP_001153019.1 | 3 - 4 | 1639 | 101 | |
NM_207406.3 | NP_997289.2 | 3 - 4 | 1639 | 101 | ||
XM_017008185.1 | XP_016863674.1 | 2 - 3 | 1296 | 101 | ||
GenBank mRNA | AB126828.1 | - | 3 - 4 | 1639 | 101 | |
AB126829.1 | - | 3 - 4 | 1639 | 101 | ||
AK125953.1 | - | 3 - 4 | 825 | 101 | ||
BC128592.1 | - | 3 - 4 | 724 | 40127911 | 101 | |
BC128593.1 | - | 3 - 4 | 724 | 40127915 | 101 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04001150.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR04015773.hg.1 | TC04001150.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04015773.hg.1 | ENST00000411720.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04015773.hg.1 | ENST00000502486.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04015773.hg.1 | ENST00000504360.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04015773.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04015773.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000611697.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000611697.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000611697.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000611697.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000611697.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000611697.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000611697.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | ENST00000611697.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400171924.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171924.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171924.hg.1 | ENST00000611697.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171924.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400171924.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400171924.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171924.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171924.hg.1 | ENST00000611697.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400171924.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400171924.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400171924.hg.1 | TC0400010553.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000502486.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000504360.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000611697.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000611697.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000611697.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000611697.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000611697.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000611697.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000611697.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | ENST00000611697.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_001159547.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_207406.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010553.hg.1 | NM_207406.3 | Refseq |
