Assay Details
Gene Symbol: BEND4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159547.1 | NP_001153019.1 | 4 - 5 | 1730 | 100 | |
NM_207406.3 | NP_997289.2 | 4 - 5 | 1730 | 100 | ||
XM_017008185.1 | XP_016863674.1 | 3 - 4 | 1387 | 100 | ||
GenBank mRNA | AB126828.1 | - | 4 - 5 | 1730 | 100 | |
AB126829.1 | - | 4 - 5 | 1730 | 100 | ||
AK125953.1 | - | 4 - 5 | 916 | 100 | ||
BC128592.1 | - | 4 - 5 | 815 | 40127911 | 100 | |
BC128593.1 | - | 4 - 5 | 815 | 40127915 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
