Assay Details
| Gene Symbol: CX3CR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171172.1 | NP_001164643.1 | 1 - 2 | 193 | 103 | |
| GenBank mRNA | DA396634.1 | - | 1 - 2 | 75 | 103 | |
| DB274136.1 | - | 1 - 2 | 193 | 103 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001304.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_001171172.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of neuroblast proliferation
microglial cell activation involved in immune response
negative regulation of chronic inflammatory response to non-antigenic stimulus
chemotaxis
cellular defense response
cell adhesion
G-protein coupled receptor signaling pathway
memory
response to wounding
viral process
negative regulation of angiogenesis
cerebral cortex cell migration
negative regulation of cell migration
positive regulation of angiogenesis
macrophage chemotaxis
chemokine-mediated signaling pathway
cellular response to lipopolysaccharide
cellular response to transforming growth factor beta stimulus
negative regulation of long-term synaptic potentiation
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
chemokine receptor activity
protein binding
C-X3-C chemokine receptor activity
C-X3-C chemokine binding
positive regulation of neuroblast proliferation
microglial cell activation involved in immune response
negative regulation of chronic inflammatory response to non-antigenic stimulus
chemotaxis
cellular defense response
cell adhesion
G-protein coupled receptor signaling pathway
memory
response to wounding
viral process
negative regulation of angiogenesis
cerebral cortex cell migration
negative regulation of cell migration
positive regulation of angiogenesis
macrophage chemotaxis
chemokine-mediated signaling pathway
cellular response to lipopolysaccharide
cellular response to transforming growth factor beta stimulus
negative regulation of long-term synaptic potentiation
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
chemokine receptor activity
protein binding
C-X3-C chemokine receptor activity
C-X3-C chemokine binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300010775.hg.1 | ENST00000358309.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010775.hg.1 | ENST00000358309.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010775.hg.1 | ENST00000358309.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010775.hg.1 | ENST00000358309.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010775.hg.1 | ENST00000358309.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010775.hg.1 | ENST00000358309.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010775.hg.1 | ENST00000399220.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010775.hg.1 | ENST00000399220.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010775.hg.1 | ENST00000399220.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010775.hg.1 | ENST00000399220.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010775.hg.1 | ENST00000399220.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010775.hg.1 | ENST00000412814.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010775.hg.1 | ENST00000412814.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010775.hg.1 | ENST00000435290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010775.hg.1 | ENST00000435290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010775.hg.1 | ENST00000435290.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010775.hg.1 | ENST00000541347.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010775.hg.1 | ENST00000541347.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010775.hg.1 | ENST00000541347.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010775.hg.1 | ENST00000541347.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010775.hg.1 | ENST00000541347.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010775.hg.1 | ENST00000542107.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010775.hg.1 | ENST00000542107.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010775.hg.1 | ENST00000542107.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010775.hg.1 | ENST00000542107.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010775.hg.1 | ENST00000542107.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010775.hg.1 | NM_001171171.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010775.hg.1 | NM_001171171.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010775.hg.1 | NM_001171171.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010775.hg.1 | NM_001171171.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010775.hg.1 | NM_001171171.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010775.hg.1 | NM_001171172.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010775.hg.1 | NM_001171172.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010775.hg.1 | NM_001171172.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010775.hg.1 | NM_001171172.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010775.hg.1 | NM_001171172.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010775.hg.1 | NM_001171174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010775.hg.1 | NM_001171174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010775.hg.1 | NM_001171174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010775.hg.1 | NM_001171174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010775.hg.1 | NM_001171174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010775.hg.1 | NM_001171174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010775.hg.1 | NM_001337.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010775.hg.1 | NM_001337.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010775.hg.1 | NM_001337.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010775.hg.1 | NM_001337.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010775.hg.1 | NM_001337.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03022480.hg.1 | TC03001304.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03022480.hg.1 | ENST00000542107.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03022480.hg.1 | NM_001171172.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300178564.hg.1 | ENST00000435290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300178564.hg.1 | ENST00000542107.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300178564.hg.1 | NM_001171172.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300178564.hg.1 | ENST00000435290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300178564.hg.1 | ENST00000542107.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300178564.hg.1 | NM_001171172.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300178564.hg.1 | TC0300010775.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300010775.hg.1 | ENST00000358309.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010775.hg.1 | ENST00000358309.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010775.hg.1 | ENST00000358309.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010775.hg.1 | ENST00000358309.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010775.hg.1 | ENST00000358309.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010775.hg.1 | ENST00000358309.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010775.hg.1 | ENST00000399220.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010775.hg.1 | ENST00000399220.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010775.hg.1 | ENST00000399220.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010775.hg.1 | ENST00000399220.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010775.hg.1 | ENST00000399220.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010775.hg.1 | ENST00000412814.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010775.hg.1 | ENST00000412814.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010775.hg.1 | ENST00000435290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010775.hg.1 | ENST00000435290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010775.hg.1 | ENST00000435290.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010775.hg.1 | ENST00000541347.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010775.hg.1 | ENST00000541347.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010775.hg.1 | ENST00000541347.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010775.hg.1 | ENST00000541347.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010775.hg.1 | ENST00000541347.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010775.hg.1 | ENST00000542107.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010775.hg.1 | ENST00000542107.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010775.hg.1 | ENST00000542107.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010775.hg.1 | ENST00000542107.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010775.hg.1 | ENST00000542107.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010775.hg.1 | NM_001171171.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010775.hg.1 | NM_001171171.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010775.hg.1 | NM_001171171.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010775.hg.1 | NM_001171171.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010775.hg.1 | NM_001171171.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010775.hg.1 | NM_001171172.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010775.hg.1 | NM_001171172.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010775.hg.1 | NM_001171172.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010775.hg.1 | NM_001171172.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010775.hg.1 | NM_001171172.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010775.hg.1 | NM_001171174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010775.hg.1 | NM_001171174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010775.hg.1 | NM_001171174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010775.hg.1 | NM_001171174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010775.hg.1 | NM_001171174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010775.hg.1 | NM_001171174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010775.hg.1 | NM_001337.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010775.hg.1 | NM_001337.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010775.hg.1 | NM_001337.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010775.hg.1 | NM_001337.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010775.hg.1 | NM_001337.3 | Refseq |

CX3CR1
Genomic Map