Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005502.hg.1 |
TC06000514.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005502.hg.1 |
ENST00000311565.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005502.hg.1 |
ENST00000337400.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005502.hg.1 |
ENST00000360694.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005502.hg.1 |
ENST00000418635.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005502.hg.1 |
ENST00000444397.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005502.hg.1 |
NM_001171818.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005502.hg.1 |
NM_001171820.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005502.hg.1 |
NM_006238.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005502.hg.1 |
NM_177435.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005506.hg.1 |
TC06000514.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005506.hg.1 |
ENST00000311565.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005506.hg.1 |
ENST00000337400.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005506.hg.1 |
ENST00000360694.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005506.hg.1 |
ENST00000418635.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005506.hg.1 |
ENST00000444397.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005506.hg.1 |
ENST00000448077.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005506.hg.1 |
ENST00000540939.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005506.hg.1 |
NM_001171818.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005506.hg.1 |
NM_001171819.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005506.hg.1 |
NM_001171820.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005506.hg.1 |
NM_006238.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005506.hg.1 |
NM_177435.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154033.hg.1 |
ENST00000311565.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154033.hg.1 |
ENST00000337400.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154033.hg.1 |
ENST00000360694.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154033.hg.1 |
ENST00000418635.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154033.hg.1 |
ENST00000448077.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154033.hg.1 |
NM_001171818.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154033.hg.1 |
NM_001171819.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154033.hg.1 |
NM_001171820.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154033.hg.1 |
NM_006238.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154033.hg.1 |
NM_177435.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154033.hg.1 |
ENST00000311565.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154033.hg.1 |
ENST00000337400.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154033.hg.1 |
ENST00000360694.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154033.hg.1 |
ENST00000418635.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154033.hg.1 |
ENST00000448077.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154033.hg.1 |
NM_001171818.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154033.hg.1 |
NM_001171819.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154033.hg.1 |
NM_001171820.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154033.hg.1 |
NM_006238.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154033.hg.1 |
NM_177435.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154033.hg.1 |
TC0600007792.hg.1 |
Clariom D TC |