Assay Details
| Gene Symbol: SETD9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171990.2 | NP_001165461.1 | 3 - 4 | 981 | 98 | |
| NM_001323018.1 | NP_001309947.1 | 3 - 4 | 830 | 98 | ||
| NM_001323019.1 | NP_001309948.1 | 3 - 4 | 981 | 98 | ||
| NM_001323020.1 | NP_001309949.1 | 3 - 4 | 830 | 98 | ||
| NM_001323022.1 | NP_001309951.1 | 3 - 4 | 558 | 98 | ||
| NM_153706.3 | NP_714917.2 | 3 - 4 | 981 | 98 | ||
| NR_136555.1 | 2 - 3 | 613 | 98 | |||
| NR_136556.1 | 2 - 3 | 613 | 98 | |||
| NR_136557.1 | 2 - 3 | 462 | 98 | |||
| NR_136558.1 | 2 - 3 | 613 | 98 | |||
| XM_005248426.4 | XP_005248483.1 | 2 - 3 | 281 | 98 | ||
| XM_011543139.2 | XP_011541441.1 | 3 - 4 | 995 | 98 | ||
| XM_011543140.2 | XP_011541442.1 | 3 - 4 | 609 | 98 | ||
| GenBank mRNA | AK309152.1 | - | 3 - 4 | 889 | 98 | |
| BC036528.1 | - | 3 - 4 | 968 | 4827793 | 98 | |
More Information
Set Membership: |
|

SETD9
Genomic Map