Assay Details
Gene Symbol: SLC25A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001636.3 | NP_001627.2 | 2 - 3 | 731 | 77 | |
GenBank mRNA | AB209822.1 | - | 2 - 3 | 686 | 77 | |
AK312336.1 | - | 2 - 3 | 698 | 77 | ||
AY007135.1 | - | 2 - 3 | 683 | 77 | ||
BC007295.1 | - | 2 - 3 | 646 | 3349670 | 77 | |
BC007850.2 | - | 2 - 3 | 685 | 4136545 | 77 | |
BC008737.2 | - | 2 - 3 | 673 | 3342722 | 77 | |
BC008935.2 | - | 2 - 3 | 678 | 2824067 | 77 | |
BC013256.1 | - | 2 - 3 | 685 | 77 | ||
BC014775.1 | - | 2 - 3 | 686 | 3458777 | 77 | |
BC031912.1 | - | 2 - 3 | 631 | 5141625 | 77 | |
BC035469.1 | - | 2 - 3 | 685 | 77 | ||
CR457400.1 | - | 2 - 3 | 594 | 77 | ||
J03592.1 | - | 1 - 2 | 486 | 77 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC0X000798.hg.1, TC0Y000115.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
translation
protein targeting to mitochondrion
apoptotic process
adenine transport
ADP transport
ATP transport
active induction of host immune response by virus
regulation of insulin secretion
transmembrane transport
structural constituent of ribosome
ATP:ADP antiporter activity
protein binding
adenine transmembrane transporter activity
translation
protein targeting to mitochondrion
apoptotic process
adenine transport
ADP transport
ATP transport
active induction of host immune response by virus
regulation of insulin secretion
transmembrane transport
structural constituent of ribosome
ATP:ADP antiporter activity
protein binding
adenine transmembrane transporter activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR0X010922.hg.1 | TC0X000798.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X010922.hg.1 | ENST00000381401.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X010922.hg.1 | ENST00000475167.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X010922.hg.1 | NM_001636.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X010918.hg.1 | TC0X000798.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X010918.hg.1 | ENST00000381401.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X010918.hg.1 | NM_001636.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X010921.hg.1 | TC0X000798.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X010921.hg.1 | ENST00000381401.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X010921.hg.1 | ENST00000475167.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X010921.hg.1 | NM_001636.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0Y001300.hg.1 | TC0Y000115.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0Y001300.hg.1 | ENST00000381401.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0Y001300.hg.1 | ENST00000475167.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0Y001300.hg.1 | NM_001636.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008908.hg.1 | ENST00000381401.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008908.hg.1 | ENST00000381401.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008908.hg.1 | ENST00000381401.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008908.hg.1 | ENST00000381401.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008908.hg.1 | ENST00000381401.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008908.hg.1 | ENST00000381401.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008908.hg.1 | ENST00000381401.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008908.hg.1 | ENST00000381401.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008908.hg.1 | ENST00000381401.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008908.hg.1 | ENST00000381401.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008908.hg.1 | ENST00000381401.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008908.hg.1 | ENST00000381401.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008908.hg.1 | ENST00000381401.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008908.hg.1 | ENST00000475167.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008908.hg.1 | ENST00000475167.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008908.hg.1 | ENST00000475167.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008908.hg.1 | ENST00000475167.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008908.hg.1 | ENST00000475167.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008908.hg.1 | ENST00000475167.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008908.hg.1 | ENST00000475167.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008908.hg.1 | ENST00000484026.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008908.hg.1 | ENST00000484026.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008908.hg.1 | ENST00000484026.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008908.hg.1 | ENST00000484026.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008908.hg.1 | ENST00000484026.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008908.hg.1 | ENST00000484026.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006882.hg.1 | ENSTR0000381401.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006882.hg.1 | ENSTR0000381401.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006882.hg.1 | ENSTR0000381401.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006882.hg.1 | ENSTR0000381401.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006882.hg.1 | ENSTR0000381401.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006882.hg.1 | ENSTR0000381401.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006882.hg.1 | ENSTR0000381401.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006882.hg.1 | ENSTR0000381401.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006882.hg.1 | ENSTR0000381401.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006882.hg.1 | ENSTR0000381401.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006882.hg.1 | ENSTR0000381401.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006882.hg.1 | ENSTR0000381401.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006882.hg.1 | ENSTR0000381401.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006882.hg.1 | ENSTR0000475167.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006882.hg.1 | ENSTR0000475167.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006882.hg.1 | ENSTR0000475167.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006882.hg.1 | ENSTR0000475167.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006882.hg.1 | ENSTR0000475167.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006882.hg.1 | ENSTR0000475167.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006882.hg.1 | ENSTR0000475167.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006882.hg.1 | ENSTR0000484026.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006882.hg.1 | ENSTR0000484026.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006882.hg.1 | ENSTR0000484026.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006882.hg.1 | ENSTR0000484026.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006882.hg.1 | ENSTR0000484026.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006882.hg.1 | ENSTR0000484026.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006882.hg.1 | NM_001636.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006882.hg.1 | NM_001636.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006882.hg.1 | NM_001636.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006882.hg.1 | NM_001636.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006882.hg.1 | NM_001636.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006882.hg.1 | NM_001636.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006882.hg.1 | NM_001636.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006882.hg.1 | NM_001636.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006882.hg.1 | NM_001636.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006882.hg.1 | NM_001636.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006882.hg.1 | NM_001636.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006882.hg.1 | NM_001636.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0Y001298.hg.1 | TC0Y000115.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0Y001298.hg.1 | ENST00000381401.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0Y001298.hg.1 | NM_001636.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0Y001299.hg.1 | TC0Y000115.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0Y001299.hg.1 | ENST00000381401.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0Y001299.hg.1 | NM_001636.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X010919.hg.1 | TC0X000798.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X010919.hg.1 | ENST00000381401.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X010919.hg.1 | NM_001636.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X010920.hg.1 | TC0X000798.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X010920.hg.1 | ENST00000381401.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X010920.hg.1 | ENST00000475167.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X010920.hg.1 | NM_001636.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900006921.hg.1 | ENST00000497974.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900006921.hg.1 | ENST00000497974.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900006921.hg.1 | ENST00000497974.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0Y00146820.hg.1 | ENSTR0000381401.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0Y00146820.hg.1 | NM_001636.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0Y00146820.hg.1 | ENSTR0000381401.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0Y00146820.hg.1 | NM_001636.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0Y00146820.hg.1 | TC0Y00006882.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0X00161198.hg.1 | ENST00000381401.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00161198.hg.1 | ENST00000475167.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00161198.hg.1 | ENST00000484026.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00161198.hg.1 | ENST00000381401.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00161198.hg.1 | ENST00000475167.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00161198.hg.1 | ENST00000484026.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00161198.hg.1 | TC0X00008908.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0Y00146821.hg.1 | ENSTR0000381401.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0Y00146821.hg.1 | ENSTR0000475167.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0Y00146821.hg.1 | NM_001636.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0Y00146821.hg.1 | ENSTR0000381401.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0Y00146821.hg.1 | ENSTR0000475167.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0Y00146821.hg.1 | NM_001636.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0Y00146821.hg.1 | TC0Y00006882.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0Y00146822.hg.1 | ENSTR0000381401.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0Y00146822.hg.1 | ENSTR0000475167.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0Y00146822.hg.1 | ENSTR0000484026.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0Y00146822.hg.1 | NM_001636.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0Y00146822.hg.1 | ENSTR0000381401.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0Y00146822.hg.1 | ENSTR0000475167.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0Y00146822.hg.1 | ENSTR0000484026.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0Y00146822.hg.1 | NM_001636.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0Y00146822.hg.1 | TC0Y00006882.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0X00161194.hg.1 | ENST00000381401.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00161194.hg.1 | ENST00000381401.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00161194.hg.1 | TC0X00008908.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0X00161197.hg.1 | ENST00000381401.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00161197.hg.1 | ENST00000475167.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00161197.hg.1 | ENST00000381401.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00161197.hg.1 | ENST00000475167.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00161197.hg.1 | TC0X00008908.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0Y00006882.hg.1 | ENSTR0000381401.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006882.hg.1 | ENSTR0000381401.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006882.hg.1 | ENSTR0000381401.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006882.hg.1 | ENSTR0000381401.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006882.hg.1 | ENSTR0000381401.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006882.hg.1 | ENSTR0000381401.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006882.hg.1 | ENSTR0000381401.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006882.hg.1 | ENSTR0000381401.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006882.hg.1 | ENSTR0000381401.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006882.hg.1 | ENSTR0000381401.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006882.hg.1 | ENSTR0000381401.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006882.hg.1 | ENSTR0000381401.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006882.hg.1 | ENSTR0000381401.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006882.hg.1 | ENSTR0000475167.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006882.hg.1 | ENSTR0000475167.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006882.hg.1 | ENSTR0000475167.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006882.hg.1 | ENSTR0000475167.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006882.hg.1 | ENSTR0000475167.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006882.hg.1 | ENSTR0000475167.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006882.hg.1 | ENSTR0000475167.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006882.hg.1 | ENSTR0000484026.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006882.hg.1 | ENSTR0000484026.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006882.hg.1 | ENSTR0000484026.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006882.hg.1 | ENSTR0000484026.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006882.hg.1 | ENSTR0000484026.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006882.hg.1 | ENSTR0000484026.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006882.hg.1 | NM_001636.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0Y00006882.hg.1 | NM_001636.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0Y00006882.hg.1 | NM_001636.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0Y00006882.hg.1 | NM_001636.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0Y00006882.hg.1 | NM_001636.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0Y00006882.hg.1 | NM_001636.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0Y00006882.hg.1 | NM_001636.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0Y00006882.hg.1 | NM_001636.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0Y00006882.hg.1 | NM_001636.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0Y00006882.hg.1 | NM_001636.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0Y00006882.hg.1 | NM_001636.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0Y00006882.hg.1 | NM_001636.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0Y00146819.hg.1 | ENSTR0000381401.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0Y00146819.hg.1 | NM_001636.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0Y00146819.hg.1 | ENSTR0000381401.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0Y00146819.hg.1 | NM_001636.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0Y00146819.hg.1 | TC0Y00006882.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0X00161196.hg.1 | ENST00000381401.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00161196.hg.1 | ENST00000381401.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00161196.hg.1 | TC0X00008908.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0900006921.hg.1 | ENST00000497974.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900006921.hg.1 | ENST00000497974.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900006921.hg.1 | ENST00000497974.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008908.hg.1 | ENST00000381401.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008908.hg.1 | ENST00000381401.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008908.hg.1 | ENST00000381401.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008908.hg.1 | ENST00000381401.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008908.hg.1 | ENST00000381401.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008908.hg.1 | ENST00000381401.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008908.hg.1 | ENST00000381401.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008908.hg.1 | ENST00000381401.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008908.hg.1 | ENST00000381401.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008908.hg.1 | ENST00000381401.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008908.hg.1 | ENST00000381401.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008908.hg.1 | ENST00000381401.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008908.hg.1 | ENST00000381401.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008908.hg.1 | ENST00000475167.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008908.hg.1 | ENST00000475167.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008908.hg.1 | ENST00000475167.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008908.hg.1 | ENST00000475167.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008908.hg.1 | ENST00000475167.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008908.hg.1 | ENST00000475167.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008908.hg.1 | ENST00000475167.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008908.hg.1 | ENST00000484026.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008908.hg.1 | ENST00000484026.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008908.hg.1 | ENST00000484026.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008908.hg.1 | ENST00000484026.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008908.hg.1 | ENST00000484026.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008908.hg.1 | ENST00000484026.5 | Ensembl |
