Assay Details
Gene Symbol: MC5R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005913.2 | NP_005904.1 | 1 | 776 | 72 | |
GenBank mRNA | BC069153.1 | - | 1 | 555 | 7217013 | 72 |
BC069545.1 | - | 1 | 554 | 7262232 | 72 | |
BC095531.1 | - | 1 | 779 | 6971958 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
