Assay Details
| Gene Symbol: PRKDC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006904.6 | NP_008835.5 | 80 - 81 | 11550 | 97 | |
| GenBank mRNA | BC037968.1 | - | 1 - 2 | 121 | 97 | |
| U35835.1 | - | 12 - 13 | 1899 | 97 | ||
| U47077.5 | - | 80 - 81 | 11550 | 97 | ||
More Information
Set Membership: |
|

PRKDC
Genomic Map