| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06016606.hg.1 |
TC06004148.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06016606.hg.1 |
ENST00000377147.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06016606.hg.1 |
ENST00000377148.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06016606.hg.1 |
ENST00000377149.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06016606.hg.1 |
NM_013937.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5001831.hg.1 |
TC6_mcf_hap5000229.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5001831.hg.1 |
NM_013937.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014254.hg.1 |
ENST00000377147.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014254.hg.1 |
ENST00000377147.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014254.hg.1 |
ENST00000377147.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014254.hg.1 |
ENST00000377147.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014254.hg.1 |
ENST00000377148.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014254.hg.1 |
ENST00000377148.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014254.hg.1 |
ENST00000377148.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014254.hg.1 |
ENST00000377148.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014254.hg.1 |
ENST00000377148.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014254.hg.1 |
ENST00000377149.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014254.hg.1 |
ENST00000377149.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014254.hg.1 |
ENST00000377149.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014254.hg.1 |
ENST00000377149.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014254.hg.1 |
ENST00000377149.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014254.hg.1 |
ENST00000377149.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014254.hg.1 |
ENST00000377149.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600014254.hg.1 |
ENST00000377149.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002031.hg.1 |
TC6_dbb_hap3000109.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002031.hg.1 |
NM_013937.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002069.hg.1 |
TC6_qbl_hap6000110.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002069.hg.1 |
NM_013937.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7001921.hg.1 |
TC6_ssto_hap7000215.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7001921.hg.1 |
NM_013937.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1000790.hg.1 |
TC6_apd_hap1000057.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1000790.hg.1 |
NM_013937.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002090.hg.1 |
TC6_cox_hap2000246.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002090.hg.1 |
NM_013937.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001611.hg.1 |
TC6_mann_hap4000212.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001611.hg.1 |
NM_013937.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600200873.hg.1 |
ENST00000377147.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600200873.hg.1 |
ENST00000377148.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600200873.hg.1 |
ENST00000377149.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600200873.hg.1 |
ENST00000377147.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600200873.hg.1 |
ENST00000377148.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600200873.hg.1 |
ENST00000377149.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600200873.hg.1 |
TC0600014254.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0600014254.hg.1 |
ENST00000377147.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014254.hg.1 |
ENST00000377147.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014254.hg.1 |
ENST00000377147.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014254.hg.1 |
ENST00000377147.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014254.hg.1 |
ENST00000377148.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014254.hg.1 |
ENST00000377148.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014254.hg.1 |
ENST00000377148.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014254.hg.1 |
ENST00000377148.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014254.hg.1 |
ENST00000377148.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014254.hg.1 |
ENST00000377149.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014254.hg.1 |
ENST00000377149.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014254.hg.1 |
ENST00000377149.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014254.hg.1 |
ENST00000377149.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014254.hg.1 |
ENST00000377149.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014254.hg.1 |
ENST00000377149.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014254.hg.1 |
ENST00000377149.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600014254.hg.1 |
ENST00000377149.5 |
Ensembl |