Assay Details
| Gene Symbol: SPEF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015417.4 | NP_056232.2 | 5 - 6 | 650 | 119 | |
| GenBank mRNA | AL040853.1 | - | 3 - 4 | 264 | 119 | |
| EF560745.1 | - | 5 - 6 | 648 | 119 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000577.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR20008498.hg.1 | ENST00000379756.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008498.hg.1 | ENST00000463490.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008498.hg.1 | NM_015417.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008498.hg.1 | TC20000577.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008499.hg.1 | ENST00000379756.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008499.hg.1 | ENST00000463490.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008499.hg.1 | NM_015417.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20008499.hg.1 | TC20000577.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008231.hg.1 | ENST00000379756.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008231.hg.1 | ENST00000379756.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008231.hg.1 | ENST00000379756.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008231.hg.1 | ENST00000379756.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008231.hg.1 | ENST00000379756.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008231.hg.1 | ENST00000379756.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008231.hg.1 | ENST00000379756.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008231.hg.1 | ENST00000379756.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008231.hg.1 | ENST00000379756.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008231.hg.1 | ENST00000379756.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008231.hg.1 | ENST00000463490.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008231.hg.1 | ENST00000463490.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008231.hg.1 | ENST00000463490.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008231.hg.1 | ENST00000463490.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008231.hg.1 | ENST00000463490.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008231.hg.1 | ENST00000463490.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008231.hg.1 | ENST00000463490.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008231.hg.1 | ENST00000463490.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008231.hg.1 | ENST00000463490.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008231.hg.1 | ENST00000463490.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008231.hg.1 | ENST00000463490.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008231.hg.1 | ENST00000463490.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008231.hg.1 | ENST00000471499.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008231.hg.1 | ENST00000471499.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008231.hg.1 | ENST00000471499.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008231.hg.1 | ENST00000471499.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008231.hg.1 | ENST00000471499.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008231.hg.1 | ENST00000471499.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008231.hg.1 | ENST00000471499.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008231.hg.1 | ENST00000471499.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008231.hg.1 | ENST00000471499.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008231.hg.1 | ENST00000471499.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008231.hg.1 | ENST00000471499.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008231.hg.1 | NM_015417.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008231.hg.1 | NM_015417.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008231.hg.1 | NM_015417.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008231.hg.1 | NM_015417.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008231.hg.1 | NM_015417.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008231.hg.1 | NM_015417.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008231.hg.1 | NM_015417.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008231.hg.1 | NM_015417.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008231.hg.1 | NM_015417.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008231.hg.1 | NM_015417.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008231.hg.1 | ENST00000379756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008231.hg.1 | ENST00000379756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008231.hg.1 | ENST00000379756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008231.hg.1 | ENST00000379756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008231.hg.1 | ENST00000379756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008231.hg.1 | ENST00000379756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008231.hg.1 | ENST00000379756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008231.hg.1 | ENST00000379756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008231.hg.1 | ENST00000379756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008231.hg.1 | ENST00000379756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008231.hg.1 | ENST00000463490.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008231.hg.1 | ENST00000463490.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008231.hg.1 | ENST00000463490.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008231.hg.1 | ENST00000463490.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008231.hg.1 | ENST00000463490.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008231.hg.1 | ENST00000463490.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008231.hg.1 | ENST00000463490.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008231.hg.1 | ENST00000463490.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008231.hg.1 | ENST00000463490.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008231.hg.1 | ENST00000463490.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008231.hg.1 | ENST00000463490.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008231.hg.1 | ENST00000463490.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008231.hg.1 | ENST00000471499.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008231.hg.1 | ENST00000471499.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008231.hg.1 | ENST00000471499.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008231.hg.1 | ENST00000471499.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008231.hg.1 | ENST00000471499.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008231.hg.1 | ENST00000471499.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008231.hg.1 | ENST00000471499.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008231.hg.1 | ENST00000471499.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008231.hg.1 | ENST00000471499.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008231.hg.1 | ENST00000471499.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008231.hg.1 | ENST00000471499.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008231.hg.1 | NM_015417.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008231.hg.1 | NM_015417.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008231.hg.1 | NM_015417.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008231.hg.1 | NM_015417.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008231.hg.1 | NM_015417.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008231.hg.1 | NM_015417.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008231.hg.1 | NM_015417.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008231.hg.1 | NM_015417.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008231.hg.1 | NM_015417.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008231.hg.1 | NM_015417.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000158105.hg.1 | ENST00000379756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000158105.hg.1 | ENST00000463490.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000158105.hg.1 | ENST00000471499.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000158105.hg.1 | NM_015417.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000158105.hg.1 | TC2000008231.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000158105.hg.1 | ENST00000379756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000158105.hg.1 | ENST00000463490.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000158105.hg.1 | ENST00000471499.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000158105.hg.1 | NM_015417.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000158106.hg.1 | ENST00000379756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000158106.hg.1 | ENST00000463490.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000158106.hg.1 | ENST00000471499.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000158106.hg.1 | NM_015417.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000158106.hg.1 | TC2000008231.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000158106.hg.1 | ENST00000379756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000158106.hg.1 | ENST00000463490.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000158106.hg.1 | ENST00000471499.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000158106.hg.1 | NM_015417.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000158107.hg.1 | ENST00000379756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000158107.hg.1 | ENST00000463490.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000158107.hg.1 | ENST00000471499.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000158107.hg.1 | NM_015417.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000158107.hg.1 | TC2000008231.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000158107.hg.1 | ENST00000379756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000158107.hg.1 | ENST00000463490.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000158107.hg.1 | ENST00000471499.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000158107.hg.1 | NM_015417.4 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 150184 | - | - |

SPEF1
Genomic Map