Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02027734.hg.1 |
TC02005023.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02027734.hg.1 |
ENST00000295065.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02027734.hg.1 |
ENST00000379383.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02027734.hg.1 |
ENST00000404530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02027734.hg.1 |
ENST00000426310.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02027734.hg.1 |
ENST00000490459.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02027734.hg.1 |
NM_001137602.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02027734.hg.1 |
NM_015955.3 |
Refseq |