Assay Details
| Gene Symbol: BDNF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143806.1 | NP_001137278.1 | 1 - 2 | 352 | 111 | |
| GenBank mRNA | AY054394.2 | - | 1 - 2 | 296 | 111 | |
| EF674518.1 | - | 1 - 2 | 352 | 111 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11001513.hg
Additional Information:
This assay is in the 5'UTR region of the AY054394.2,EF674518.1,NM_001143806.1 transcripts.
Set Membership: |
|

BDNF
Genomic Map