Assay Details
Gene Symbol: GPR75-ASB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164165.1 | NP_001157637.1 | 1 - 2 | 206 | 85 | |
GenBank mRNA | BC110915.1 | - | 1 - 2 | 128 | 5764788 | 85 |
DC314536.1 | - | 1 - 2 | 336 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200016667.hg.1 | ENST00000406625.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016667.hg.1 | ENST00000406625.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016667.hg.1 | ENST00000406625.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016667.hg.1 | ENST00000406625.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016667.hg.1 | ENST00000406625.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016667.hg.1 | ENST00000406625.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016667.hg.1 | ENST00000406625.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016667.hg.1 | ENST00000406625.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016667.hg.1 | ENST00000406625.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016667.hg.1 | ENST00000406625.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016667.hg.1 | ENST00000406625.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016667.hg.1 | ENST00000406625.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016667.hg.1 | ENST00000406625.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016667.hg.1 | ENST00000406625.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016667.hg.1 | ENST00000406625.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016667.hg.1 | ENST00000406625.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016667.hg.1 | ENST00000406625.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016667.hg.1 | ENST00000459916.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016667.hg.1 | ENST00000459916.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016667.hg.1 | ENST00000459916.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016667.hg.1 | ENST00000459916.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016667.hg.1 | ENST00000459916.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016667.hg.1 | ENST00000498475.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016667.hg.1 | ENST00000498475.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016667.hg.1 | ENST00000498475.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016667.hg.1 | ENST00000498475.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016667.hg.1 | ENST00000498475.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016667.hg.1 | ENST00000498475.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016667.hg.1 | ENST00000498475.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016667.hg.1 | NM_001164165.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016667.hg.1 | NM_001164165.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016667.hg.1 | NM_001164165.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016667.hg.1 | NM_001164165.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016667.hg.1 | NM_001164165.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016667.hg.1 | NM_001164165.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016667.hg.1 | NM_001164165.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016667.hg.1 | NM_001164165.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016667.hg.1 | NM_001164165.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016667.hg.1 | NM_001164165.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016667.hg.1 | NM_001164165.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016667.hg.1 | NM_001164165.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016667.hg.1 | NM_001164165.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016667.hg.1 | NM_001164165.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016667.hg.1 | NM_001164165.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016667.hg.1 | NM_001164165.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016668.hg.1 | ENST00000394705.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016668.hg.1 | ENST00000394705.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016668.hg.1 | ENST00000394705.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016668.hg.1 | ENST00000394705.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016668.hg.1 | ENST00000394705.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016668.hg.1 | ENST00000394705.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016668.hg.1 | ENST00000394705.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016668.hg.1 | NM_006794.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016668.hg.1 | NM_006794.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016668.hg.1 | NM_006794.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016668.hg.1 | NM_006794.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016668.hg.1 | NM_006794.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016668.hg.1 | NM_006794.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016668.hg.1 | NM_006794.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016668.hg.1 | NM_006794.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200226305.hg.1 | ENST00000406625.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200226305.hg.1 | ENST00000498475.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200226305.hg.1 | NM_001164165.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200226305.hg.1 | ENST00000406625.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200226305.hg.1 | ENST00000498475.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200226305.hg.1 | NM_001164165.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200226305.hg.1 | TC0200016667.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200226306.hg.1 | ENST00000406625.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200226306.hg.1 | ENST00000498475.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200226306.hg.1 | NM_001164165.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200226306.hg.1 | ENST00000406625.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200226306.hg.1 | ENST00000498475.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200226306.hg.1 | NM_001164165.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200226306.hg.1 | TC0200016667.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0200016667.hg.1 | ENST00000406625.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016667.hg.1 | ENST00000406625.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016667.hg.1 | ENST00000406625.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016667.hg.1 | ENST00000406625.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016667.hg.1 | ENST00000406625.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016667.hg.1 | ENST00000406625.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016667.hg.1 | ENST00000406625.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016667.hg.1 | ENST00000406625.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016667.hg.1 | ENST00000406625.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016667.hg.1 | ENST00000406625.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016667.hg.1 | ENST00000406625.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016667.hg.1 | ENST00000406625.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016667.hg.1 | ENST00000406625.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016667.hg.1 | ENST00000406625.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016667.hg.1 | ENST00000406625.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016667.hg.1 | ENST00000406625.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016667.hg.1 | ENST00000406625.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016667.hg.1 | ENST00000459916.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016667.hg.1 | ENST00000459916.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016667.hg.1 | ENST00000459916.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016667.hg.1 | ENST00000459916.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016667.hg.1 | ENST00000459916.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016667.hg.1 | ENST00000498475.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016667.hg.1 | ENST00000498475.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016667.hg.1 | ENST00000498475.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016667.hg.1 | ENST00000498475.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016667.hg.1 | ENST00000498475.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016667.hg.1 | ENST00000498475.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016667.hg.1 | ENST00000498475.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016667.hg.1 | NM_001164165.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016667.hg.1 | NM_001164165.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016667.hg.1 | NM_001164165.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016667.hg.1 | NM_001164165.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016667.hg.1 | NM_001164165.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016667.hg.1 | NM_001164165.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016667.hg.1 | NM_001164165.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016667.hg.1 | NM_001164165.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016667.hg.1 | NM_001164165.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016667.hg.1 | NM_001164165.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016667.hg.1 | NM_001164165.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016667.hg.1 | NM_001164165.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016667.hg.1 | NM_001164165.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016667.hg.1 | NM_001164165.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016667.hg.1 | NM_001164165.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016667.hg.1 | NM_001164165.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016668.hg.1 | ENST00000394705.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016668.hg.1 | ENST00000394705.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016668.hg.1 | ENST00000394705.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016668.hg.1 | ENST00000394705.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016668.hg.1 | ENST00000394705.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016668.hg.1 | ENST00000394705.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016668.hg.1 | ENST00000394705.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016668.hg.1 | NM_006794.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016668.hg.1 | NM_006794.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016668.hg.1 | NM_006794.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016668.hg.1 | NM_006794.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016668.hg.1 | NM_006794.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016668.hg.1 | NM_006794.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016668.hg.1 | NM_006794.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016668.hg.1 | NM_006794.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200226301.hg.1 | ENST00000406625.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200226301.hg.1 | ENST00000459916.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200226301.hg.1 | NM_001164165.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200226301.hg.1 | ENST00000406625.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200226301.hg.1 | ENST00000459916.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200226301.hg.1 | NM_001164165.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200226301.hg.1 | TC0200016667.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200226298.hg.1 | ENST00000406625.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200226298.hg.1 | NM_001164165.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200226298.hg.1 | ENST00000406625.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200226298.hg.1 | NM_001164165.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200226298.hg.1 | TC0200016667.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200226299.hg.1 | ENST00000406625.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200226299.hg.1 | ENST00000459916.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200226299.hg.1 | NM_001164165.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200226299.hg.1 | ENST00000406625.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200226299.hg.1 | ENST00000459916.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200226299.hg.1 | NM_001164165.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200226299.hg.1 | TC0200016667.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200226303.hg.1 | ENST00000406625.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200226303.hg.1 | ENST00000459916.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200226303.hg.1 | ENST00000498475.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200226303.hg.1 | NM_001164165.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200226303.hg.1 | ENST00000406625.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200226303.hg.1 | ENST00000459916.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200226303.hg.1 | ENST00000498475.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200226303.hg.1 | NM_001164165.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200226303.hg.1 | TC0200016667.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200226307.hg.1 | ENST00000406625.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200226307.hg.1 | NM_001164165.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200226307.hg.1 | ENST00000406625.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200226307.hg.1 | NM_001164165.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200226307.hg.1 | TC0200016667.hg.1 | Clariom D TC |
