Assay Details
| Gene Symbol: PXMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018663.2 | NP_061133.1 | 1 - 2 | 187 | 64 | |
| GenBank mRNA | AF250136.1 | - | 1 - 2 | 187 | 64 | |
| AY044439.1 | - | 1 - 2 | 122 | 64 | ||
| BC009836.1 | - | 4 - 5 | 64 | |||
| BC073997.1 | - | 1 - 2 | 158 | 6171609 | 64 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12003266.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200012730.hg.1 | ENST00000317479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012730.hg.1 | ENST00000317479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012730.hg.1 | ENST00000317479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012730.hg.1 | ENST00000317479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012730.hg.1 | ENST00000317479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012730.hg.1 | ENST00000317479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012730.hg.1 | ENST00000317479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012730.hg.1 | ENST00000317479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012730.hg.1 | ENST00000317479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012730.hg.1 | ENST00000317479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012730.hg.1 | ENST00000317479.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012730.hg.1 | ENST00000428960.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012730.hg.1 | ENST00000428960.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012730.hg.1 | ENST00000428960.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012730.hg.1 | ENST00000428960.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012730.hg.1 | ENST00000428960.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012730.hg.1 | ENST00000428960.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012730.hg.1 | ENST00000428960.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012730.hg.1 | ENST00000454379.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012730.hg.1 | ENST00000454379.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012730.hg.1 | ENST00000454379.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012730.hg.1 | ENST00000539093.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012730.hg.1 | ENST00000539093.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012730.hg.1 | ENST00000539093.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012730.hg.1 | ENST00000539093.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012730.hg.1 | ENST00000539093.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012730.hg.1 | ENST00000543589.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012730.hg.1 | ENST00000543589.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012730.hg.1 | ENST00000543589.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012730.hg.1 | ENST00000543589.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012730.hg.1 | ENST00000543589.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012730.hg.1 | ENST00000543589.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012730.hg.1 | ENST00000543589.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012730.hg.1 | NM_018663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012730.hg.1 | NM_018663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012730.hg.1 | NM_018663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012730.hg.1 | NM_018663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012730.hg.1 | NM_018663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012730.hg.1 | NM_018663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012730.hg.1 | NM_018663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012730.hg.1 | NM_018663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012730.hg.1 | NM_018663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012730.hg.1 | NM_018663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012730.hg.1 | NM_018663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12014029.hg.1 | TC12003266.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12014029.hg.1 | ENST00000317479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12014029.hg.1 | ENST00000543589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12014029.hg.1 | NM_018663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12014027.hg.1 | TC12003266.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12014027.hg.1 | ENST00000317479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12014027.hg.1 | NM_018663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12014033.hg.1 | TC12003266.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12014033.hg.1 | ENST00000317479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12014033.hg.1 | ENST00000539093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12014033.hg.1 | ENST00000543589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12014033.hg.1 | NM_018663.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200202111.hg.1 | ENST00000317479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200202111.hg.1 | ENST00000539093.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200202111.hg.1 | ENST00000543589.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200202111.hg.1 | NM_018663.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200202111.hg.1 | ENST00000317479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200202111.hg.1 | ENST00000539093.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200202111.hg.1 | ENST00000543589.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200202111.hg.1 | NM_018663.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200202111.hg.1 | TC1200012730.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200012730.hg.1 | ENST00000317479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012730.hg.1 | ENST00000317479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012730.hg.1 | ENST00000317479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012730.hg.1 | ENST00000317479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012730.hg.1 | ENST00000317479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012730.hg.1 | ENST00000317479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012730.hg.1 | ENST00000317479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012730.hg.1 | ENST00000317479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012730.hg.1 | ENST00000317479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012730.hg.1 | ENST00000317479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012730.hg.1 | ENST00000317479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012730.hg.1 | ENST00000428960.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012730.hg.1 | ENST00000428960.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012730.hg.1 | ENST00000428960.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012730.hg.1 | ENST00000428960.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012730.hg.1 | ENST00000428960.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012730.hg.1 | ENST00000428960.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012730.hg.1 | ENST00000428960.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012730.hg.1 | ENST00000454379.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012730.hg.1 | ENST00000454379.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012730.hg.1 | ENST00000454379.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012730.hg.1 | ENST00000539093.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012730.hg.1 | ENST00000539093.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012730.hg.1 | ENST00000539093.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012730.hg.1 | ENST00000539093.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012730.hg.1 | ENST00000539093.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012730.hg.1 | ENST00000543589.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012730.hg.1 | ENST00000543589.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012730.hg.1 | ENST00000543589.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012730.hg.1 | ENST00000543589.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012730.hg.1 | ENST00000543589.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012730.hg.1 | ENST00000543589.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012730.hg.1 | ENST00000543589.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012730.hg.1 | NM_018663.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012730.hg.1 | NM_018663.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012730.hg.1 | NM_018663.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012730.hg.1 | NM_018663.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012730.hg.1 | NM_018663.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012730.hg.1 | NM_018663.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012730.hg.1 | NM_018663.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012730.hg.1 | NM_018663.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012730.hg.1 | NM_018663.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012730.hg.1 | NM_018663.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012730.hg.1 | NM_018663.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200202107.hg.1 | ENST00000317479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200202107.hg.1 | NM_018663.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200202107.hg.1 | ENST00000317479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200202107.hg.1 | NM_018663.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200202107.hg.1 | TC1200012730.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200202110.hg.1 | ENST00000317479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200202110.hg.1 | ENST00000543589.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200202110.hg.1 | NM_018663.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200202110.hg.1 | ENST00000317479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200202110.hg.1 | ENST00000543589.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200202110.hg.1 | NM_018663.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200202110.hg.1 | TC1200012730.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200202109.hg.1 | ENST00000317479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200202109.hg.1 | ENST00000543589.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200202109.hg.1 | NM_018663.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200202109.hg.1 | ENST00000317479.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200202109.hg.1 | ENST00000543589.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200202109.hg.1 | NM_018663.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200202109.hg.1 | TC1200012730.hg.1 | Clariom D TC |

PXMP2
Genomic Map