Assay Details
| Gene Symbol: PCDHAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018898.3 | NP_061721.2 | 1 - 2 | 2611 | 113 | |
| GenBank mRNA | AF152303.1 | - | 1 - 2 | 2435 | 113 | |
More Information
Set Membership: |
|

PCDHAC1
Genomic Map