Assay Details
| Gene Symbol: COL8A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001850.4 | NP_001841.2 | 3 - 4 | 259 | 100 | |
| NM_020351.3 | NP_065084.2 | 2 - 3 | 197 | 100 | ||
| GenBank mRNA | AY550974.1 | - | 1 - 2 | 57 | 100 | |
| BC013581.1 | - | 3 - 4 | 237 | 3875911 | 100 | |
| CA393523.1 | - | 2 - 3 | 183 | 100 | ||
| CN484310.1 | - | 2 - 3 | 183 | 100 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000502.hg.1
Additional Information:
This assay is in the 5'UTR region of the AY550974.1,BC013581.1,NM_001850.4,NM_020351.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | ENST00000261037.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | ENST00000261037.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | ENST00000261037.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | ENST00000261037.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | ENST00000261037.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | ENST00000261037.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | ENST00000261037.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | ENST00000261037.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | ENST00000261037.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | ENST00000261037.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | ENST00000273342.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | ENST00000273342.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | ENST00000273342.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | ENST00000273342.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | ENST00000273342.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | ENST00000273342.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | ENST00000273342.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | ENST00000452013.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | ENST00000452013.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | ENST00000452013.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | ENST00000452013.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | ENST00000452013.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | ENST00000462604.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | ENST00000462604.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | ENST00000463753.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | ENST00000463753.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | ENST00000463753.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | ENST00000463753.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | ENST00000463753.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | ENST00000474648.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | ENST00000474648.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | ENST00000474648.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | ENST00000474648.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | ENST00000474648.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | ENST00000474648.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | ENST00000483969.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | ENST00000483969.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | ENST00000483969.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | ENST00000483969.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | ENST00000483969.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | ENST00000483969.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | ENST00000621757.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | ENST00000621757.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | ENST00000621757.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | ENST00000621757.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | ENST00000621757.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | ENST00000621757.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | NM_001850.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | NM_001850.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | NM_001850.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | NM_001850.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | NM_001850.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | NM_001850.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | NM_001850.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | NM_001850.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | NM_020351.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | NM_020351.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | NM_020351.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | NM_020351.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | NM_020351.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | NM_020351.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008130.hg.1 | NM_020351.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009430.hg.1 | TC03000502.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009430.hg.1 | ENST00000261037.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009430.hg.1 | ENST00000273342.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009430.hg.1 | NM_001850.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009430.hg.1 | NM_020351.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009429.hg.1 | TC03000502.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009429.hg.1 | ENST00000261037.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009429.hg.1 | ENST00000273342.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009429.hg.1 | NM_001850.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009429.hg.1 | NM_020351.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300157906.hg.1 | ENST00000261037.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157906.hg.1 | ENST00000273342.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157906.hg.1 | ENST00000452013.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157906.hg.1 | ENST00000621757.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157906.hg.1 | NM_001850.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300157906.hg.1 | NM_020351.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300157906.hg.1 | ENST00000261037.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157906.hg.1 | ENST00000273342.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157906.hg.1 | ENST00000452013.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157906.hg.1 | ENST00000621757.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157906.hg.1 | NM_001850.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300157906.hg.1 | NM_020351.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300157906.hg.1 | TC0300008130.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300157908.hg.1 | ENST00000261037.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157908.hg.1 | ENST00000273342.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157908.hg.1 | ENST00000452013.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157908.hg.1 | ENST00000621757.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157908.hg.1 | NM_001850.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300157908.hg.1 | NM_020351.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300157908.hg.1 | ENST00000261037.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157908.hg.1 | ENST00000273342.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157908.hg.1 | ENST00000452013.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157908.hg.1 | ENST00000621757.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157908.hg.1 | NM_001850.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300157908.hg.1 | NM_020351.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300157908.hg.1 | TC0300008130.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | ENST00000261037.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | ENST00000261037.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | ENST00000261037.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | ENST00000261037.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | ENST00000261037.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | ENST00000261037.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | ENST00000261037.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | ENST00000261037.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | ENST00000261037.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | ENST00000261037.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | ENST00000273342.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | ENST00000273342.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | ENST00000273342.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | ENST00000273342.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | ENST00000273342.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | ENST00000273342.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | ENST00000273342.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | ENST00000452013.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | ENST00000452013.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | ENST00000452013.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | ENST00000452013.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | ENST00000452013.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | ENST00000462604.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | ENST00000462604.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | ENST00000463753.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | ENST00000463753.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | ENST00000463753.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | ENST00000463753.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | ENST00000463753.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | ENST00000474648.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | ENST00000474648.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | ENST00000474648.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | ENST00000474648.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | ENST00000474648.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | ENST00000474648.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | ENST00000483969.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | ENST00000483969.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | ENST00000483969.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | ENST00000483969.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | ENST00000483969.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | ENST00000483969.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | ENST00000621757.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | ENST00000621757.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | ENST00000621757.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | ENST00000621757.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | ENST00000621757.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | ENST00000621757.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | NM_001850.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | NM_001850.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | NM_001850.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | NM_001850.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | NM_001850.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | NM_001850.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | NM_001850.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | NM_001850.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | NM_020351.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | NM_020351.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | NM_020351.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | NM_020351.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | NM_020351.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | NM_020351.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008130.hg.1 | NM_020351.3 | Refseq |

COL8A1
Genomic Map