Assay Details
| Gene Symbol: TRAPPC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166621.1 | NP_001160093.1 | 2 - 3 | 157 | 65 | |
| NM_021210.4 | NP_067033.1 | 1 - 2 | 217 | 65 | ||
| GenBank mRNA | AK314038.1 | - | 1 - 2 | 223 | 65 | |
| BC032717.1 | - | 1 - 2 | 219 | 5518603 | 65 | |
| BI457771.1 | - | 1 - 2 | 219 | 65 | ||
| BI600633.1 | - | 2 - 3 | 162 | 65 | ||
| BM762006.1 | - | 2 - 3 | 131 | 65 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17001098.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000303731.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000303731.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000303731.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000303731.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000303731.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000303731.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000303731.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000303731.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000540486.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000540486.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000540486.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000540486.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000540486.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000540486.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000540486.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000540486.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000571739.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000571739.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000571739.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000571739.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000571739.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000571739.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000571947.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000571947.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000571947.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000571947.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000572656.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000572656.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000572656.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000572656.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000572656.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000573989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000573989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000573989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000573989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000575639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000575639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000575639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000575639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000575639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | ENST00000575639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NM_001166621.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NM_001166621.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NM_001166621.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NM_001166621.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NM_001166621.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NM_001166621.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NM_001166621.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NM_001166621.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NM_021210.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NM_021210.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NM_021210.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NM_021210.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NM_021210.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NM_021210.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NM_021210.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NM_021210.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NR_030684.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NR_030684.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NR_030684.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NR_030684.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NR_030684.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NR_030684.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NR_030684.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NR_030697.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NR_030697.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NR_030697.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NR_030697.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NR_030697.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NR_030697.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012352.hg.1 | NR_030697.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17014777.hg.1 | ENST00000303731.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17014777.hg.1 | ENST00000540486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17014777.hg.1 | NM_001166621.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17014777.hg.1 | NM_021210.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17014777.hg.1 | TC17001098.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1700205175.hg.1 | ENST00000303731.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205175.hg.1 | ENST00000540486.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205175.hg.1 | ENST00000571947.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205175.hg.1 | ENST00000572656.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205175.hg.1 | ENST00000573989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205175.hg.1 | NM_001166621.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700205175.hg.1 | NM_021210.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700205175.hg.1 | ENST00000303731.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205175.hg.1 | ENST00000540486.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205175.hg.1 | ENST00000571947.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205175.hg.1 | ENST00000572656.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205175.hg.1 | ENST00000573989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205175.hg.1 | NM_001166621.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700205175.hg.1 | NM_021210.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700205175.hg.1 | TC1700012352.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000303731.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000303731.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000303731.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000303731.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000303731.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000303731.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000303731.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000303731.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000540486.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000540486.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000540486.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000540486.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000540486.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000540486.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000540486.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000540486.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000571739.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000571739.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000571739.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000571739.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000571739.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000571739.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000571947.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000571947.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000571947.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000571947.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000572656.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000572656.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000572656.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000572656.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000572656.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000573989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000573989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000573989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000573989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000575639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000575639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000575639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000575639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000575639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | ENST00000575639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NM_001166621.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NM_001166621.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NM_001166621.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NM_001166621.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NM_001166621.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NM_001166621.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NM_001166621.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NM_001166621.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NM_021210.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NM_021210.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NM_021210.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NM_021210.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NM_021210.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NM_021210.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NM_021210.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NM_021210.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NR_030684.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NR_030684.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NR_030684.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NR_030684.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NR_030684.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NR_030684.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NR_030684.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NR_030697.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NR_030697.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NR_030697.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NR_030697.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NR_030697.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NR_030697.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012352.hg.1 | NR_030697.1 | Refseq |

TRAPPC1
Genomic Map