Assay Details
| Gene Symbol: VKORC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024006.5 | NP_076869.1 | 2 - 3 | 556 | 76 | |
| GenBank mRNA | AK312005.1 | - | 2 - 3 | 469 | 76 | |
| AY423044.1 | - | 2 - 3 | 286 | 76 | ||
| AY466113.1 | - | 2 - 3 | 498 | 76 | ||
| AY521634.1 | - | 2 - 3 | 286 | 76 | ||
| BC000828.1 | - | 2 - 3 | 286 | 76 | ||
| BC002911.2 | - | 2 - 3 | 324 | 3944530 | 76 | |
| BI822140.1 | - | 2 - 3 | 512 | 76 | ||
| BM663008.1 | - | 1 - 2 | 513 | 76 | ||
| KU178762.1 | - | 2 - 3 | 286 | 76 | ||
More Information
Set Membership: |
|

VKORC1
Genomic Map