Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017901.hg.1 |
ENST00000372873.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017901.hg.1 |
NM_024875.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017901.hg.1 |
TC10001414.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011029.hg.1 |
ENST00000372873.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011029.hg.1 |
ENST00000372873.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011029.hg.1 |
ENST00000372873.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011029.hg.1 |
ENST00000372873.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011029.hg.1 |
ENST00000394810.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011029.hg.1 |
ENST00000394810.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011029.hg.1 |
ENST00000394810.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011029.hg.1 |
ENST00000394810.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011029.hg.1 |
ENST00000394810.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011029.hg.1 |
ENST00000394810.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011029.hg.1 |
ENST00000394810.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011029.hg.1 |
ENST00000394810.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011029.hg.1 |
ENST00000606523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011029.hg.1 |
ENST00000606523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011029.hg.1 |
ENST00000606523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011029.hg.1 |
NM_001114133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011029.hg.1 |
NM_001114133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011029.hg.1 |
NM_001114133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011029.hg.1 |
NM_001114133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011029.hg.1 |
NM_001114133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011029.hg.1 |
NM_001114133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011029.hg.1 |
NM_001114133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011029.hg.1 |
NM_001114133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011029.hg.1 |
NM_001114133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011029.hg.1 |
NM_024875.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011029.hg.1 |
NM_024875.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011029.hg.1 |
NM_024875.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011029.hg.1 |
NM_024875.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011029.hg.1 |
ENST00000372873.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011029.hg.1 |
ENST00000372873.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011029.hg.1 |
ENST00000372873.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011029.hg.1 |
ENST00000372873.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011029.hg.1 |
ENST00000394810.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011029.hg.1 |
ENST00000394810.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011029.hg.1 |
ENST00000394810.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011029.hg.1 |
ENST00000394810.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011029.hg.1 |
ENST00000394810.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011029.hg.1 |
ENST00000394810.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011029.hg.1 |
ENST00000394810.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011029.hg.1 |
ENST00000394810.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011029.hg.1 |
ENST00000606523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011029.hg.1 |
ENST00000606523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011029.hg.1 |
ENST00000606523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011029.hg.1 |
NM_001114133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011029.hg.1 |
NM_001114133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011029.hg.1 |
NM_001114133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011029.hg.1 |
NM_001114133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011029.hg.1 |
NM_001114133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011029.hg.1 |
NM_001114133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011029.hg.1 |
NM_001114133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011029.hg.1 |
NM_001114133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011029.hg.1 |
NM_001114133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011029.hg.1 |
NM_024875.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011029.hg.1 |
NM_024875.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011029.hg.1 |
NM_024875.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011029.hg.1 |
NM_024875.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000175188.hg.1 |
ENST00000372873.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175188.hg.1 |
NM_024875.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000175188.hg.1 |
ENST00000372873.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175188.hg.1 |
NM_024875.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000175188.hg.1 |
TC1000011029.hg.1 |
Clariom D TC |