Assay Details
| Gene Symbol: ZNF394 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032164.2 | NP_115540.2 | 2 - 3 | 739 | 71 | |
| GenBank mRNA | AK022360.1 | - | 2 - 3 | 782 | 71 | |
| AK172865.1 | - | 3 - 4 | 660 | 71 | ||
| AK222948.1 | - | 2 - 3 | 732 | 71 | ||
| AY642122.1 | - | 2 - 3 | 766 | 71 | ||
| BC025241.1 | - | 2 - 3 | 766 | 4641268 | 71 | |
| BC051847.1 | - | 2 - 3 | 739 | 6007172 | 71 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | ENST00000337673.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | ENST00000337673.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | ENST00000337673.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | ENST00000337673.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | ENST00000337673.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | ENST00000337673.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | ENST00000337673.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | ENST00000337673.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | ENST00000337673.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | ENST00000337673.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | ENST00000394177.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | ENST00000394177.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | ENST00000394177.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | ENST00000394177.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | ENST00000394177.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | ENST00000394177.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | ENST00000394177.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | ENST00000426306.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | ENST00000426306.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | ENST00000426306.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | ENST00000426306.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | ENST00000426306.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | ENST00000426306.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | ENST00000426306.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | ENST00000426306.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | ENST00000462024.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | ENST00000462024.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | ENST00000464401.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | ENST00000464401.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | ENST00000464401.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | ENST00000464401.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | ENST00000481881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | ENST00000481881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | ENST00000481881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | ENST00000481881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | ENST00000481881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | ENST00000481881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | ENST00000481881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | ENST00000481881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | ENST00000481881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | ENST00000481881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | ENST00000485576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | ENST00000485576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | ENST00000485576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | ENST00000485576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | ENST00000485576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | ENST00000485576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | ENST00000485576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | ENST00000485576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | ENST00000485576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | NM_032164.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | NM_032164.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | NM_032164.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | NM_032164.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | NM_032164.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | NM_032164.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | NM_032164.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | NM_032164.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | NM_032164.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011944.hg.1 | NM_032164.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | ENST00000337673.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | ENST00000337673.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | ENST00000337673.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | ENST00000337673.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | ENST00000337673.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | ENST00000337673.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | ENST00000337673.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | ENST00000337673.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | ENST00000337673.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | ENST00000337673.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | ENST00000394177.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | ENST00000394177.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | ENST00000394177.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | ENST00000394177.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | ENST00000394177.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | ENST00000394177.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | ENST00000394177.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | ENST00000426306.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | ENST00000426306.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | ENST00000426306.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | ENST00000426306.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | ENST00000426306.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | ENST00000426306.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | ENST00000426306.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | ENST00000426306.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | ENST00000462024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | ENST00000462024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | ENST00000464401.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | ENST00000464401.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | ENST00000464401.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | ENST00000464401.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | ENST00000481881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | ENST00000481881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | ENST00000481881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | ENST00000481881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | ENST00000481881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | ENST00000481881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | ENST00000481881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | ENST00000481881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | ENST00000481881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | ENST00000481881.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | ENST00000485576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | ENST00000485576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | ENST00000485576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | ENST00000485576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | ENST00000485576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | ENST00000485576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | ENST00000485576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | ENST00000485576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | ENST00000485576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | NM_032164.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | NM_032164.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | NM_032164.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | NM_032164.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | NM_032164.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | NM_032164.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | NM_032164.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | NM_032164.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | NM_032164.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011944.hg.1 | NM_032164.3 | Refseq |

ZNF394
Genomic Map