Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014330.hg.1 |
ENST00000395106.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014330.hg.1 |
ENST00000555633.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014330.hg.1 |
NM_032753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014330.hg.1 |
TC19001050.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014332.hg.1 |
ENST00000395106.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014332.hg.1 |
ENST00000555633.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014332.hg.1 |
NM_032753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014332.hg.1 |
TC19001050.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009308.hg.1 |
ENST00000555633.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009308.hg.1 |
ENST00000555633.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009308.hg.1 |
ENST00000555633.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009308.hg.1 |
ENST00000555633.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009308.hg.1 |
ENST00000555633.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009308.hg.1 |
ENST00000555633.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009308.hg.1 |
ENST00000555633.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009308.hg.1 |
ENST00000555978.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009308.hg.1 |
ENST00000555978.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009308.hg.1 |
ENST00000555978.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009308.hg.1 |
ENST00000555978.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009308.hg.1 |
ENST00000555978.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009308.hg.1 |
ENST00000555978.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009308.hg.1 |
NM_032753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009308.hg.1 |
NM_032753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009308.hg.1 |
NM_032753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009308.hg.1 |
NM_032753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009308.hg.1 |
NM_032753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009308.hg.1 |
NM_032753.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900174232.hg.1 |
ENST00000555633.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900174232.hg.1 |
ENST00000555978.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900174232.hg.1 |
NM_032753.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900174232.hg.1 |
TC1900009308.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900174232.hg.1 |
ENST00000555633.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900174232.hg.1 |
ENST00000555978.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900174232.hg.1 |
NM_032753.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009308.hg.1 |
ENST00000555633.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009308.hg.1 |
ENST00000555633.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009308.hg.1 |
ENST00000555633.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009308.hg.1 |
ENST00000555633.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009308.hg.1 |
ENST00000555633.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009308.hg.1 |
ENST00000555633.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009308.hg.1 |
ENST00000555633.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009308.hg.1 |
ENST00000555978.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009308.hg.1 |
ENST00000555978.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009308.hg.1 |
ENST00000555978.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009308.hg.1 |
ENST00000555978.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009308.hg.1 |
ENST00000555978.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009308.hg.1 |
ENST00000555978.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009308.hg.1 |
NM_032753.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009308.hg.1 |
NM_032753.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009308.hg.1 |
NM_032753.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009308.hg.1 |
NM_032753.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009308.hg.1 |
NM_032753.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009308.hg.1 |
NM_032753.3 |
Refseq |