Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12007294.hg.1 |
ENST00000266643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12007294.hg.1 |
NM_138396.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12007294.hg.1 |
TC12000543.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12007293.hg.1 |
ENST00000266643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12007293.hg.1 |
NM_138396.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12007293.hg.1 |
TC12000543.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007899.hg.1 |
ENST00000266643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007899.hg.1 |
ENST00000266643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007899.hg.1 |
ENST00000266643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007899.hg.1 |
ENST00000266643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007899.hg.1 |
ENST00000266643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007899.hg.1 |
ENST00000266643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007899.hg.1 |
ENST00000266643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007899.hg.1 |
ENST00000266643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007899.hg.1 |
ENST00000548358.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007899.hg.1 |
ENST00000548358.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007899.hg.1 |
ENST00000548358.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007899.hg.1 |
ENST00000548358.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007899.hg.1 |
ENST00000548358.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007899.hg.1 |
ENST00000548358.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007899.hg.1 |
ENST00000552279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007899.hg.1 |
ENST00000552279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007899.hg.1 |
ENST00000552279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007899.hg.1 |
ENST00000552279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007899.hg.1 |
NM_138396.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007899.hg.1 |
NM_138396.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007899.hg.1 |
NM_138396.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007899.hg.1 |
NM_138396.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007899.hg.1 |
NM_138396.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007899.hg.1 |
NM_138396.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007899.hg.1 |
NM_138396.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007899.hg.1 |
NM_138396.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200157926.hg.1 |
ENST00000266643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200157926.hg.1 |
ENST00000552279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200157926.hg.1 |
NM_138396.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200157926.hg.1 |
ENST00000266643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200157926.hg.1 |
ENST00000552279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200157926.hg.1 |
NM_138396.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200157926.hg.1 |
TC1200007899.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200157927.hg.1 |
ENST00000266643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200157927.hg.1 |
ENST00000552279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200157927.hg.1 |
NM_138396.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200157927.hg.1 |
ENST00000266643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200157927.hg.1 |
ENST00000552279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200157927.hg.1 |
NM_138396.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200157927.hg.1 |
TC1200007899.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200007899.hg.1 |
ENST00000266643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007899.hg.1 |
ENST00000266643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007899.hg.1 |
ENST00000266643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007899.hg.1 |
ENST00000266643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007899.hg.1 |
ENST00000266643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007899.hg.1 |
ENST00000266643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007899.hg.1 |
ENST00000266643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007899.hg.1 |
ENST00000266643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007899.hg.1 |
ENST00000548358.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007899.hg.1 |
ENST00000548358.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007899.hg.1 |
ENST00000548358.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007899.hg.1 |
ENST00000548358.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007899.hg.1 |
ENST00000548358.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007899.hg.1 |
ENST00000548358.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007899.hg.1 |
ENST00000552279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007899.hg.1 |
ENST00000552279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007899.hg.1 |
ENST00000552279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007899.hg.1 |
ENST00000552279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007899.hg.1 |
NM_138396.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007899.hg.1 |
NM_138396.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007899.hg.1 |
NM_138396.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007899.hg.1 |
NM_138396.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007899.hg.1 |
NM_138396.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007899.hg.1 |
NM_138396.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007899.hg.1 |
NM_138396.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007899.hg.1 |
NM_138396.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200157924.hg.1 |
ENST00000266643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200157924.hg.1 |
NM_138396.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200157924.hg.1 |
ENST00000266643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200157924.hg.1 |
NM_138396.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200157924.hg.1 |
TC1200007899.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200157925.hg.1 |
ENST00000266643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200157925.hg.1 |
NM_138396.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200157925.hg.1 |
ENST00000266643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200157925.hg.1 |
NM_138396.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200157925.hg.1 |
TC1200007899.hg.1 |
Clariom D TC |