Assay Details
| Gene Symbol: LRRC45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144999.3 | NP_659436.1 | 2 - 3 | 622 | 87 | |
| GenBank mRNA | BC014109.2 | - | 2 - 3 | 520 | 4330269 | 87 |
| BI837975.1 | - | 2 - 3 | 591 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000306688.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000306688.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000306688.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000306688.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000306688.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000306688.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000306688.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000306688.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000306688.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000306688.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000306688.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000306688.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000306688.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000306688.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000306688.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000306688.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000306688.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000306688.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000306688.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000306688.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000306688.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000306688.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000577638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000577638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000577638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000577638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000577638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000581227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000581227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000581227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000581227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000581227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000581227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000581227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000581227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000581227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000581227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000581227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000581227.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000582083.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000582083.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000582083.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000582083.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000583302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000583302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000583302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000583302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000583302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000583302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000583302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000583302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000583302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000583302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000583302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000583302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000583302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000583383.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000583383.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000583383.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000583383.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000583383.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | ENST00000583383.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | NM_144999.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | NM_144999.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | NM_144999.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | NM_144999.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | NM_144999.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | NM_144999.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | NM_144999.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | NM_144999.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | NM_144999.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | NM_144999.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | NM_144999.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | NM_144999.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | NM_144999.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | NM_144999.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | NM_144999.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | NM_144999.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | NM_144999.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | NM_144999.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | NM_144999.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | NM_144999.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | NM_144999.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009217.hg.1 | NM_144999.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169969.hg.1 | ENST00000306688.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169969.hg.1 | ENST00000577638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169969.hg.1 | ENST00000583383.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169969.hg.1 | NM_144999.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169969.hg.1 | ENST00000306688.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169969.hg.1 | ENST00000577638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169969.hg.1 | ENST00000583383.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700169969.hg.1 | NM_144999.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700169969.hg.1 | TC1700009217.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000306688.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000306688.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000306688.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000306688.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000306688.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000306688.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000306688.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000306688.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000306688.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000306688.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000306688.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000306688.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000306688.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000306688.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000306688.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000306688.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000306688.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000306688.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000306688.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000306688.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000306688.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000306688.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000577638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000577638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000577638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000577638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000577638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000581227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000581227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000581227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000581227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000581227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000581227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000581227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000581227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000581227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000581227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000581227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000581227.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000582083.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000582083.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000582083.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000582083.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000583302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000583302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000583302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000583302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000583302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000583302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000583302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000583302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000583302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000583302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000583302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000583302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000583302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000583383.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000583383.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000583383.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000583383.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000583383.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | ENST00000583383.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | NM_144999.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | NM_144999.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | NM_144999.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | NM_144999.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | NM_144999.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | NM_144999.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | NM_144999.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | NM_144999.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | NM_144999.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | NM_144999.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | NM_144999.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | NM_144999.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | NM_144999.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | NM_144999.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | NM_144999.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | NM_144999.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | NM_144999.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | NM_144999.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | NM_144999.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | NM_144999.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | NM_144999.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009217.hg.1 | NM_144999.3 | Refseq |

LRRC45
Genomic Map