Assay Details
| Gene Symbol: DIRAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145173.3 | NP_660156.1 | 1 - 2 | 92 | 54 | |
| GenBank mRNA | BC030660.1 | - | 1 - 2 | 71 | 4814337 | 54 |
| DA754806.1 | - | 1 - 2 | 92 | 54 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001034.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC030660.1,NM_145173.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19014102.hg.1 | ENST00000323469.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014102.hg.1 | NM_145173.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014102.hg.1 | TC19001034.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014105.hg.1 | ENST00000323469.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014105.hg.1 | NM_145173.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014105.hg.1 | TC19001034.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011842.hg.1 | ENST00000323469.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011842.hg.1 | ENST00000323469.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011842.hg.1 | ENST00000323469.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011842.hg.1 | ENST00000323469.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011842.hg.1 | ENST00000323469.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011842.hg.1 | ENST00000323469.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011842.hg.1 | ENST00000323469.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011842.hg.1 | ENST00000323469.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011842.hg.1 | ENST00000323469.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011842.hg.1 | ENST00000585334.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011842.hg.1 | ENST00000585334.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011842.hg.1 | ENST00000585334.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011842.hg.1 | ENST00000585334.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011842.hg.1 | ENST00000585334.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011842.hg.1 | ENST00000588128.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011842.hg.1 | ENST00000588128.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011842.hg.1 | ENST00000588128.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011842.hg.1 | ENST00000588128.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011842.hg.1 | NM_145173.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011842.hg.1 | NM_145173.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011842.hg.1 | NM_145173.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011842.hg.1 | NM_145173.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011842.hg.1 | NM_145173.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011842.hg.1 | NM_145173.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011842.hg.1 | NM_145173.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011842.hg.1 | NM_145173.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014104.hg.1 | ENST00000323469.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014104.hg.1 | NM_145173.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014104.hg.1 | TC19001034.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014106.hg.1 | ENST00000323469.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014106.hg.1 | NM_145173.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014106.hg.1 | TC19001034.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014103.hg.1 | ENST00000323469.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014103.hg.1 | NM_145173.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014103.hg.1 | TC19001034.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1900203553.hg.1 | ENST00000323469.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203553.hg.1 | ENST00000585334.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203553.hg.1 | ENST00000588128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203553.hg.1 | NM_145173.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900203553.hg.1 | TC1900011842.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900203553.hg.1 | ENST00000323469.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203553.hg.1 | ENST00000585334.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203553.hg.1 | ENST00000588128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203553.hg.1 | NM_145173.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900203551.hg.1 | ENST00000323469.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203551.hg.1 | ENST00000585334.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203551.hg.1 | NM_145173.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900203551.hg.1 | TC1900011842.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900203551.hg.1 | ENST00000323469.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203551.hg.1 | ENST00000585334.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203551.hg.1 | NM_145173.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900203556.hg.1 | ENST00000323469.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203556.hg.1 | ENST00000588128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203556.hg.1 | NM_145173.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900203556.hg.1 | TC1900011842.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900203556.hg.1 | ENST00000323469.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203556.hg.1 | ENST00000588128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203556.hg.1 | NM_145173.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900203558.hg.1 | ENST00000323469.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203558.hg.1 | NM_145173.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900203558.hg.1 | TC1900011842.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900203558.hg.1 | ENST00000323469.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203558.hg.1 | NM_145173.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900203549.hg.1 | ENST00000323469.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203549.hg.1 | NM_145173.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900203549.hg.1 | TC1900011842.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900203549.hg.1 | ENST00000323469.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203549.hg.1 | NM_145173.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900203552.hg.1 | ENST00000323469.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203552.hg.1 | ENST00000585334.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203552.hg.1 | ENST00000588128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203552.hg.1 | NM_145173.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900203552.hg.1 | TC1900011842.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900203552.hg.1 | ENST00000323469.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203552.hg.1 | ENST00000585334.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203552.hg.1 | ENST00000588128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203552.hg.1 | NM_145173.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900203550.hg.1 | ENST00000323469.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203550.hg.1 | ENST00000585334.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203550.hg.1 | NM_145173.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900203550.hg.1 | TC1900011842.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900203550.hg.1 | ENST00000323469.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203550.hg.1 | ENST00000585334.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203550.hg.1 | NM_145173.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900203557.hg.1 | ENST00000323469.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203557.hg.1 | NM_145173.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900203557.hg.1 | TC1900011842.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900203557.hg.1 | ENST00000323469.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203557.hg.1 | NM_145173.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011842.hg.1 | ENST00000323469.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011842.hg.1 | ENST00000323469.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011842.hg.1 | ENST00000323469.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011842.hg.1 | ENST00000323469.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011842.hg.1 | ENST00000323469.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011842.hg.1 | ENST00000323469.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011842.hg.1 | ENST00000323469.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011842.hg.1 | ENST00000323469.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011842.hg.1 | ENST00000323469.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011842.hg.1 | ENST00000585334.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011842.hg.1 | ENST00000585334.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011842.hg.1 | ENST00000585334.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011842.hg.1 | ENST00000585334.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011842.hg.1 | ENST00000585334.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011842.hg.1 | ENST00000588128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011842.hg.1 | ENST00000588128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011842.hg.1 | ENST00000588128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011842.hg.1 | ENST00000588128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011842.hg.1 | NM_145173.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011842.hg.1 | NM_145173.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011842.hg.1 | NM_145173.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011842.hg.1 | NM_145173.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011842.hg.1 | NM_145173.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011842.hg.1 | NM_145173.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011842.hg.1 | NM_145173.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011842.hg.1 | NM_145173.3 | Refseq |

DIRAS1
Genomic Map