Assay Details
Gene Symbol: TMEM196 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152774.3 | NP_689987.3 | 1 - 2 | 229 | 79 | |
XM_011515249.2 | XP_011513551.1 | 1 - 2 | 1148 | 79 | ||
XM_017011929.1 | XP_016867418.1 | 1 - 2 | 1148 | 79 | ||
GenBank mRNA | BX647256.1 | - | 1 - 2 | 841 | 79 | |
BX648384.1 | - | 1 - 2 | 229 | 79 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC07001179.hg.1
Additional Information:
This assay is in the 5'UTR region of the BX647256.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR07018387.hg.1 | TC07001179.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07018387.hg.1 | ENST00000405764.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07018387.hg.1 | ENST00000405844.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07018387.hg.1 | ENST00000422233.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07018387.hg.1 | ENST00000433641.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07018387.hg.1 | ENST00000493519.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07018387.hg.1 | NM_152774.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010398.hg.1 | ENST00000405764.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010398.hg.1 | ENST00000405764.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010398.hg.1 | ENST00000405764.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010398.hg.1 | ENST00000405764.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010398.hg.1 | ENST00000405764.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010398.hg.1 | ENST00000405764.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010398.hg.1 | ENST00000405764.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010398.hg.1 | ENST00000405764.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010398.hg.1 | ENST00000405764.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010398.hg.1 | ENST00000405764.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010398.hg.1 | ENST00000405764.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010398.hg.1 | ENST00000405844.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010398.hg.1 | ENST00000405844.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010398.hg.1 | ENST00000405844.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010398.hg.1 | ENST00000405844.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010398.hg.1 | ENST00000405844.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010398.hg.1 | ENST00000405844.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010398.hg.1 | ENST00000405844.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010398.hg.1 | ENST00000405844.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010398.hg.1 | ENST00000405844.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010398.hg.1 | ENST00000405844.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010398.hg.1 | ENST00000405844.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010398.hg.1 | ENST00000405844.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010398.hg.1 | ENST00000422233.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010398.hg.1 | ENST00000422233.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010398.hg.1 | ENST00000422233.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010398.hg.1 | ENST00000422233.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010398.hg.1 | ENST00000422233.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010398.hg.1 | ENST00000422233.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010398.hg.1 | ENST00000422233.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010398.hg.1 | ENST00000433641.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010398.hg.1 | ENST00000433641.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010398.hg.1 | ENST00000433641.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010398.hg.1 | ENST00000433641.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010398.hg.1 | ENST00000433641.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010398.hg.1 | ENST00000433641.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010398.hg.1 | ENST00000433641.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010398.hg.1 | ENST00000493519.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010398.hg.1 | ENST00000493519.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010398.hg.1 | ENST00000493519.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010398.hg.1 | ENST00000493519.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010398.hg.1 | ENST00000493519.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010398.hg.1 | NM_152774.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010398.hg.1 | NM_152774.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010398.hg.1 | NM_152774.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010398.hg.1 | NM_152774.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010398.hg.1 | NM_152774.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010398.hg.1 | NM_152774.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010398.hg.1 | NM_152774.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010398.hg.1 | NM_152774.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07018388.hg.1 | TC07001179.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07018388.hg.1 | ENST00000405764.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07018388.hg.1 | ENST00000405844.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07018388.hg.1 | NM_152774.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700173931.hg.1 | ENST00000405764.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700173931.hg.1 | ENST00000405844.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700173931.hg.1 | NM_152774.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700173931.hg.1 | ENST00000405764.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700173931.hg.1 | ENST00000405844.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700173931.hg.1 | NM_152774.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700173931.hg.1 | TC0700010398.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0700010398.hg.1 | ENST00000405764.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010398.hg.1 | ENST00000405764.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010398.hg.1 | ENST00000405764.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010398.hg.1 | ENST00000405764.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010398.hg.1 | ENST00000405764.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010398.hg.1 | ENST00000405764.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010398.hg.1 | ENST00000405764.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010398.hg.1 | ENST00000405764.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010398.hg.1 | ENST00000405764.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010398.hg.1 | ENST00000405764.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010398.hg.1 | ENST00000405764.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010398.hg.1 | ENST00000405844.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010398.hg.1 | ENST00000405844.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010398.hg.1 | ENST00000405844.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010398.hg.1 | ENST00000405844.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010398.hg.1 | ENST00000405844.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010398.hg.1 | ENST00000405844.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010398.hg.1 | ENST00000405844.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010398.hg.1 | ENST00000405844.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010398.hg.1 | ENST00000405844.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010398.hg.1 | ENST00000405844.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010398.hg.1 | ENST00000405844.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010398.hg.1 | ENST00000405844.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010398.hg.1 | ENST00000422233.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010398.hg.1 | ENST00000422233.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010398.hg.1 | ENST00000422233.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010398.hg.1 | ENST00000422233.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010398.hg.1 | ENST00000422233.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010398.hg.1 | ENST00000422233.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010398.hg.1 | ENST00000422233.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010398.hg.1 | ENST00000433641.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010398.hg.1 | ENST00000433641.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010398.hg.1 | ENST00000433641.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010398.hg.1 | ENST00000433641.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010398.hg.1 | ENST00000433641.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010398.hg.1 | ENST00000433641.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010398.hg.1 | ENST00000433641.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010398.hg.1 | ENST00000493519.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010398.hg.1 | ENST00000493519.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010398.hg.1 | ENST00000493519.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010398.hg.1 | ENST00000493519.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010398.hg.1 | ENST00000493519.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010398.hg.1 | NM_152774.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010398.hg.1 | NM_152774.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010398.hg.1 | NM_152774.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010398.hg.1 | NM_152774.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010398.hg.1 | NM_152774.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010398.hg.1 | NM_152774.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010398.hg.1 | NM_152774.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010398.hg.1 | NM_152774.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700173929.hg.1 | ENST00000405764.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700173929.hg.1 | ENST00000405844.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700173929.hg.1 | ENST00000422233.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700173929.hg.1 | ENST00000433641.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700173929.hg.1 | ENST00000493519.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700173929.hg.1 | NM_152774.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700173929.hg.1 | ENST00000405764.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700173929.hg.1 | ENST00000405844.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700173929.hg.1 | ENST00000422233.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700173929.hg.1 | ENST00000433641.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700173929.hg.1 | ENST00000493519.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700173929.hg.1 | NM_152774.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700173929.hg.1 | TC0700010398.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700173930.hg.1 | ENST00000405764.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700173930.hg.1 | ENST00000405844.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700173930.hg.1 | NM_152774.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700173930.hg.1 | ENST00000405764.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700173930.hg.1 | ENST00000405844.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700173930.hg.1 | NM_152774.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700173930.hg.1 | TC0700010398.hg.1 | Clariom D TC |
