Assay Details
| Gene Symbol: FAM89B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152832.2 | NP_690045.1 | 1 - 2 | 610 | 97 | |
| GenBank mRNA | BC032373.1 | - | 1 - 2 | 621 | 5123262 | 97 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11003462.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11007897.hg.1 | TC11003462.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11007897.hg.1 | ENST00000316409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11007897.hg.1 | ENST00000377088.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11007897.hg.1 | ENST00000449319.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11007897.hg.1 | ENST00000530349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11007897.hg.1 | NM_001098784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11007897.hg.1 | NM_001098785.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11007897.hg.1 | NM_152832.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11007898.hg.1 | TC11003462.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11007898.hg.1 | ENST00000316409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11007898.hg.1 | ENST00000449319.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11007898.hg.1 | ENST00000530349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11007898.hg.1 | NM_001098784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11007898.hg.1 | NM_001098785.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11007898.hg.1 | NM_152832.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013043.hg.1 | ENST00000316409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013043.hg.1 | ENST00000316409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013043.hg.1 | ENST00000316409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013043.hg.1 | ENST00000316409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013043.hg.1 | ENST00000316409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013043.hg.1 | ENST00000316409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013043.hg.1 | ENST00000316409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013043.hg.1 | ENST00000449319.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013043.hg.1 | ENST00000449319.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013043.hg.1 | ENST00000449319.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013043.hg.1 | ENST00000449319.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013043.hg.1 | ENST00000449319.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013043.hg.1 | ENST00000449319.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013043.hg.1 | ENST00000449319.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013043.hg.1 | ENST00000530349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013043.hg.1 | ENST00000530349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013043.hg.1 | ENST00000530349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013043.hg.1 | ENST00000530349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013043.hg.1 | ENST00000530349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013043.hg.1 | ENST00000530349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013043.hg.1 | NM_001098784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013043.hg.1 | NM_001098784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013043.hg.1 | NM_001098784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013043.hg.1 | NM_001098784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013043.hg.1 | NM_001098784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013043.hg.1 | NM_001098784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013043.hg.1 | NM_001098784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013043.hg.1 | NM_001098784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013043.hg.1 | NM_001098785.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013043.hg.1 | NM_001098785.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013043.hg.1 | NM_001098785.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013043.hg.1 | NM_001098785.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013043.hg.1 | NM_001098785.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013043.hg.1 | NM_001098785.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013043.hg.1 | NM_001098785.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013043.hg.1 | NM_001098785.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013043.hg.1 | NM_152832.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013043.hg.1 | NM_152832.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013043.hg.1 | NM_152832.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013043.hg.1 | NM_152832.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013043.hg.1 | NM_152832.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013043.hg.1 | NM_152832.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013043.hg.1 | NM_152832.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100201361.hg.1 | ENST00000316409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100201361.hg.1 | ENST00000449319.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100201361.hg.1 | ENST00000530349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100201361.hg.1 | NM_001098784.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100201361.hg.1 | NM_001098785.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100201361.hg.1 | NM_152832.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100201361.hg.1 | ENST00000316409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100201361.hg.1 | ENST00000449319.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100201361.hg.1 | ENST00000530349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100201361.hg.1 | NM_001098784.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100201361.hg.1 | NM_001098785.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100201361.hg.1 | NM_152832.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100201361.hg.1 | TC1100013043.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100013043.hg.1 | ENST00000316409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013043.hg.1 | ENST00000316409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013043.hg.1 | ENST00000316409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013043.hg.1 | ENST00000316409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013043.hg.1 | ENST00000316409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013043.hg.1 | ENST00000316409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013043.hg.1 | ENST00000316409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013043.hg.1 | ENST00000449319.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013043.hg.1 | ENST00000449319.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013043.hg.1 | ENST00000449319.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013043.hg.1 | ENST00000449319.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013043.hg.1 | ENST00000449319.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013043.hg.1 | ENST00000449319.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013043.hg.1 | ENST00000449319.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013043.hg.1 | ENST00000530349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013043.hg.1 | ENST00000530349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013043.hg.1 | ENST00000530349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013043.hg.1 | ENST00000530349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013043.hg.1 | ENST00000530349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013043.hg.1 | ENST00000530349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013043.hg.1 | NM_001098784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013043.hg.1 | NM_001098784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013043.hg.1 | NM_001098784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013043.hg.1 | NM_001098784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013043.hg.1 | NM_001098784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013043.hg.1 | NM_001098784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013043.hg.1 | NM_001098784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013043.hg.1 | NM_001098784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013043.hg.1 | NM_001098785.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013043.hg.1 | NM_001098785.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013043.hg.1 | NM_001098785.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013043.hg.1 | NM_001098785.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013043.hg.1 | NM_001098785.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013043.hg.1 | NM_001098785.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013043.hg.1 | NM_001098785.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013043.hg.1 | NM_001098785.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013043.hg.1 | NM_152832.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013043.hg.1 | NM_152832.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013043.hg.1 | NM_152832.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013043.hg.1 | NM_152832.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013043.hg.1 | NM_152832.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013043.hg.1 | NM_152832.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013043.hg.1 | NM_152832.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100201359.hg.1 | ENST00000316409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100201359.hg.1 | ENST00000449319.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100201359.hg.1 | ENST00000530349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100201359.hg.1 | NM_001098784.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100201359.hg.1 | NM_001098785.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100201359.hg.1 | NM_152832.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100201359.hg.1 | ENST00000316409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100201359.hg.1 | ENST00000449319.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100201359.hg.1 | ENST00000530349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100201359.hg.1 | NM_001098784.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100201359.hg.1 | NM_001098785.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100201359.hg.1 | NM_152832.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100201359.hg.1 | TC1100013043.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100201360.hg.1 | ENST00000316409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100201360.hg.1 | ENST00000449319.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100201360.hg.1 | ENST00000530349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100201360.hg.1 | NM_001098784.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100201360.hg.1 | NM_001098785.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100201360.hg.1 | NM_152832.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100201360.hg.1 | ENST00000316409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100201360.hg.1 | ENST00000449319.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100201360.hg.1 | ENST00000530349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100201360.hg.1 | NM_001098784.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100201360.hg.1 | NM_001098785.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100201360.hg.1 | NM_152832.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100201360.hg.1 | TC1100013043.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 221314 | - | - |

FAM89B
Genomic Map