Assay Details
| Gene Symbol: SPATA31D4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145197.1 | NP_001138669.1 | 3 - 4 | 381 | 112 | |
| GenBank mRNA | AK125847.1 | - | 3 - 4 | 326 | 112 | |
| AK308327.1 | - | 3 - 4 | 322 | 112 | ||
| Gene Symbol: SPATA31D3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207416.2 | NP_997299.2 | 3 - 4 | 381 | 112 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09000366.hg.1, TC09000367.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 256191 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 232185 | AK129489.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 138077 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 128337 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 305767 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 186525 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 353203 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 174775 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3216819 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 20434 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3216819 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3216819 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8156101 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8156100 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8156099 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8156102 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8156098 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007709.hg.1 | ENST00000376459.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007709.hg.1 | ENST00000376459.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007709.hg.1 | ENST00000376459.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007709.hg.1 | ENST00000376459.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007709.hg.1 | ENST00000527857.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007709.hg.1 | ENST00000527857.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007709.hg.1 | ENST00000527857.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007709.hg.1 | ENST00000527857.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007709.hg.1 | ENST00000527857.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007709.hg.1 | ENST00000527857.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007709.hg.1 | NR_026851.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007709.hg.1 | NR_026851.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007709.hg.1 | NR_026851.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007709.hg.1 | NR_026851.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007709.hg.1 | NR_026851.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007709.hg.1 | NR_026851.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004021.hg.1 | TC09000367.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004021.hg.1 | ENST00000334208.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004021.hg.1 | NM_207416.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007710.hg.1 | ENST00000419782.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007710.hg.1 | ENST00000419782.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007710.hg.1 | ENST00000419782.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007710.hg.1 | ENST00000419782.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007710.hg.1 | NM_001145197.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007710.hg.1 | NM_001145197.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007710.hg.1 | NM_001145197.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007710.hg.1 | NM_001145197.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007710.hg.1 | NM_001145197.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007710.hg.1 | NM_001145197.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012145.hg.1 | ENST00000445385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012145.hg.1 | ENST00000445385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012145.hg.1 | ENST00000445385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012145.hg.1 | ENST00000445385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012145.hg.1 | NM_207416.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012145.hg.1 | NM_207416.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012145.hg.1 | NM_207416.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012145.hg.1 | NM_207416.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012145.hg.1 | NM_207416.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012145.hg.1 | NM_207416.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004016.hg.1 | TC09000366.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004016.hg.1 | ENST00000341875.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004016.hg.1 | NM_001145197.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010552.hg.1 | ENST00000427387.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010552.hg.1 | ENST00000427387.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010552.hg.1 | ENST00000585776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010552.hg.1 | ENST00000585776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010552.hg.1 | ENST00000585776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010552.hg.1 | ENST00000585776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010552.hg.1 | ENST00000585776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010552.hg.1 | ENST00000585776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010552.hg.1 | ENST00000585776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010552.hg.1 | ENST00000585776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010552.hg.1 | ENST00000585776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010552.hg.1 | ENST00000585776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010552.hg.1 | ENST00000585776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010552.hg.1 | ENST00000585776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010552.hg.1 | ENST00000585776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010552.hg.1 | ENST00000585776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010552.hg.1 | ENST00000585776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010552.hg.1 | ENST00000585776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010552.hg.1 | ENST00000585776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010552.hg.1 | ENST00000585776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010552.hg.1 | ENST00000585776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010552.hg.1 | ENST00000585776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010552.hg.1 | ENST00000592744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010552.hg.1 | ENST00000592744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010552.hg.1 | ENST00000592744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010552.hg.1 | ENST00000592744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900184191.hg.1 | ENST00000445385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900184191.hg.1 | NM_207416.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900184191.hg.1 | ENST00000445385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900184191.hg.1 | NM_207416.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900184191.hg.1 | TC0900012145.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900007709.hg.1 | ENST00000376459.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007709.hg.1 | ENST00000376459.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007709.hg.1 | ENST00000376459.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007709.hg.1 | ENST00000376459.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007709.hg.1 | ENST00000527857.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007709.hg.1 | ENST00000527857.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007709.hg.1 | ENST00000527857.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007709.hg.1 | ENST00000527857.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007709.hg.1 | ENST00000527857.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007709.hg.1 | ENST00000527857.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007709.hg.1 | NR_026851.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007709.hg.1 | NR_026851.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007709.hg.1 | NR_026851.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007709.hg.1 | NR_026851.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007709.hg.1 | NR_026851.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007709.hg.1 | NR_026851.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010552.hg.1 | ENST00000427387.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010552.hg.1 | ENST00000427387.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010552.hg.1 | ENST00000585776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010552.hg.1 | ENST00000585776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010552.hg.1 | ENST00000585776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010552.hg.1 | ENST00000585776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010552.hg.1 | ENST00000585776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010552.hg.1 | ENST00000585776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010552.hg.1 | ENST00000585776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010552.hg.1 | ENST00000585776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010552.hg.1 | ENST00000585776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010552.hg.1 | ENST00000585776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010552.hg.1 | ENST00000585776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010552.hg.1 | ENST00000585776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010552.hg.1 | ENST00000585776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010552.hg.1 | ENST00000585776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010552.hg.1 | ENST00000585776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010552.hg.1 | ENST00000585776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010552.hg.1 | ENST00000585776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010552.hg.1 | ENST00000585776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010552.hg.1 | ENST00000585776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010552.hg.1 | ENST00000585776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010552.hg.1 | ENST00000592744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010552.hg.1 | ENST00000592744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010552.hg.1 | ENST00000592744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010552.hg.1 | ENST00000592744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012145.hg.1 | ENST00000445385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012145.hg.1 | ENST00000445385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012145.hg.1 | ENST00000445385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012145.hg.1 | ENST00000445385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012145.hg.1 | NM_207416.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012145.hg.1 | NM_207416.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012145.hg.1 | NM_207416.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012145.hg.1 | NM_207416.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012145.hg.1 | NM_207416.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012145.hg.1 | NM_207416.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900152109.hg.1 | NM_001145197.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900152109.hg.1 | NM_001145197.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900152109.hg.1 | TC0900007710.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900007710.hg.1 | ENST00000419782.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007710.hg.1 | ENST00000419782.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007710.hg.1 | ENST00000419782.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007710.hg.1 | ENST00000419782.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007710.hg.1 | NM_001145197.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007710.hg.1 | NM_001145197.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007710.hg.1 | NM_001145197.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007710.hg.1 | NM_001145197.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007710.hg.1 | NM_001145197.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007710.hg.1 | NM_001145197.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900152108.hg.1 | ENST00000419782.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900152108.hg.1 | NM_001145197.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900152108.hg.1 | ENST00000419782.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900152108.hg.1 | NM_001145197.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900152108.hg.1 | TC0900007710.hg.1 | Clariom D TC |
| Human Genome U95 Set | 42860_at | - | - |
| Human Genome U95 Set | 42860_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A038837 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A038837 | - | - |

SPATA31D4
Genomic Map