Assay Details
Gene Symbol: ZNF880 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145434.1 | NP_001138906.1 | 1 - 2 | 62 | 92 | |
XM_017026812.1 | XP_016882301.1 | 1 - 2 | 79 | 92 | ||
GenBank mRNA | BC058819.1 | - | 1 - 2 | 43 | 92 | |
DC419753.1 | - | 1 - 2 | 62 | 92 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC19000797.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC058819.1,NM_001145434.1,XM_017026812.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR19011212.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19011212.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19011212.hg.1 | TC19000797.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR19011204.hg.1 | ENST00000344085.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19011204.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19011204.hg.1 | ENST00000424032.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19011204.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19011204.hg.1 | TC19000797.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000344085.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000344085.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000344085.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000344085.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000344085.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000424032.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000424032.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000424032.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000424032.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000424032.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000424032.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000595099.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000595099.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000597976.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000597976.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000597976.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000597976.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000597976.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000597976.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000600321.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000600321.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000600321.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000600321.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000600321.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | ENST00000600321.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008696.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19011206.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19011206.hg.1 | ENST00000424032.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19011206.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19011206.hg.1 | TC19000797.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR19011203.hg.1 | ENST00000344085.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19011203.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19011203.hg.1 | ENST00000424032.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19011203.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19011203.hg.1 | TC19000797.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR19011209.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19011209.hg.1 | ENST00000424032.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19011209.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19011209.hg.1 | TC19000797.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR19011210.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19011210.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19011210.hg.1 | TC19000797.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1900008698.hg.1 | ENST00000620439.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19011211.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19011211.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19011211.hg.1 | TC19000797.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR19011205.hg.1 | ENST00000344085.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19011205.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19011205.hg.1 | ENST00000424032.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19011205.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19011205.hg.1 | TC19000797.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1900167985.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167985.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900167985.hg.1 | TC1900008696.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900167985.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167985.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008698.hg.1 | ENST00000620439.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167977.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167977.hg.1 | ENST00000424032.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167977.hg.1 | ENST00000597976.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167977.hg.1 | ENST00000600321.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167977.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900167977.hg.1 | TC1900008696.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900167977.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167977.hg.1 | ENST00000424032.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167977.hg.1 | ENST00000597976.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167977.hg.1 | ENST00000600321.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167977.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900167975.hg.1 | ENST00000344085.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167975.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167975.hg.1 | ENST00000424032.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167975.hg.1 | ENST00000597976.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167975.hg.1 | ENST00000600321.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167975.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900167975.hg.1 | TC1900008696.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900167975.hg.1 | ENST00000344085.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167975.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167975.hg.1 | ENST00000424032.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167975.hg.1 | ENST00000597976.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167975.hg.1 | ENST00000600321.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167975.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900167987.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167987.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900167987.hg.1 | TC1900008696.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900167987.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167987.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900167984.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167984.hg.1 | ENST00000424032.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167984.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900167984.hg.1 | TC1900008696.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900167984.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167984.hg.1 | ENST00000424032.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167984.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900167988.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167988.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900167988.hg.1 | TC1900008696.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900167988.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167988.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900167974.hg.1 | ENST00000344085.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167974.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167974.hg.1 | ENST00000424032.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167974.hg.1 | ENST00000595099.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167974.hg.1 | ENST00000597976.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167974.hg.1 | ENST00000600321.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167974.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900167974.hg.1 | TC1900008696.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900167974.hg.1 | ENST00000344085.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167974.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167974.hg.1 | ENST00000424032.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167974.hg.1 | ENST00000595099.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167974.hg.1 | ENST00000597976.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167974.hg.1 | ENST00000600321.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167974.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900167976.hg.1 | ENST00000344085.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167976.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167976.hg.1 | ENST00000424032.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167976.hg.1 | ENST00000597976.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167976.hg.1 | ENST00000600321.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167976.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900167976.hg.1 | TC1900008696.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900167976.hg.1 | ENST00000344085.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167976.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167976.hg.1 | ENST00000424032.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167976.hg.1 | ENST00000597976.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167976.hg.1 | ENST00000600321.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900167976.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000344085.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000344085.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000344085.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000344085.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000344085.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000422689.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000424032.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000424032.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000424032.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000424032.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000424032.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000424032.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000595099.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000595099.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000597976.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000597976.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000597976.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000597976.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000597976.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000597976.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000600321.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000600321.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000600321.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000600321.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000600321.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | ENST00000600321.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | NM_001145434.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008696.hg.1 | NM_001145434.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 129307 | - | - |
