Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.0 ST Array |
NM_014513_at |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8031372 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8031296 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8039853 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8039857 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8039834 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8039893 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8031364 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8031299 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8031373 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8039877 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8039904 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8039844 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8039849 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8031271 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8031363 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8039898 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8031292 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8039895 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8039899 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8031272 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8031371 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8031301 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8031310 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8039879 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8031300 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8039851 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8039882 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8039883 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8031284 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8039848 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8039837 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8039855 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8031362 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8031360 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8039881 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8031365 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8039900 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8039852 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8039901 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8031286 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8039894 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8039836 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8031273 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8031288 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8039856 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8031369 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8031307 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8039858 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8031291 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8031283 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8039850 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8031294 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8039833 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8039897 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8031287 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8039892 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8031358 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8039896 |
- |
- |
Affymetrix HT HG-U133+ PM Array Plate |
208203_PM_x_at |
- |
- |
Affymetrix Human Clariom D Assay |
PSR1900169417.hg.1 |
ENST00000339924.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169417.hg.1 |
NM_014513.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900169417.hg.1 |
TC1900008879.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900169417.hg.1 |
ENST00000339924.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900169417.hg.1 |
NM_014513.2 |
Refseq |
Human Genome U133 Plus 2.0 Array |
217318_x_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
217318_PM_x_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
208203_PM_x_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
208203_x_at |
- |
- |
Human Genome U133A 2.0 Array |
208203_PM_x_at |
- |
- |
Human Genome U133A 2.0 Array |
208203_x_at |
- |
- |
Human Genome U133A 2.0 Array |
217318_x_at |
- |
- |
Human Genome U133A 2.0 Array |
217318_PM_x_at |
- |
- |
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) |
NM_014513_at |
- |
- |
AffymetrixPathwork Diagnostics Pathchip |
pw520574 |
- |
- |
Human Genome U95 Set |
36886_f_at |
- |
- |
Human Genome U95 Set |
36886_PM_f_at |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
208203_x_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
208203_x_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
208203_x_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
208203_x_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
208203_x_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
208203_x_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
208203_x_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
208203_x_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
208203_x_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
208203_x_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
208203_x_at9 |
- |
- |
Human Genome U133 Set |
217318_PM_x_at |
- |
- |
Human Genome U133 Set |
208203_x_at |
- |
- |
Human Genome U133 Set |
217318_x_at |
- |
- |
Human Genome U133 Set |
208203_PM_x_at |
- |
- |