Assay Details
Gene Symbol: GEMIN8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042480.1 | NP_001035945.1 | 1 - 2 | 141 | 114 | |
XM_017029617.1 | XP_016885106.1 | 1 - 2 | 312 | 114 | ||
XM_017029618.1 | XP_016885107.1 | 1 - 2 | 312 | 114 | ||
GenBank mRNA | BC014324.1 | - | 1 - 2 | 122 | 3930643 | 114 |
BC020785.1 | - | 1 - 2 | 118 | 4808527 | 114 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC0X000870.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC014324.1,BC020785.1,NM_001042480.1,XM_017029617.1,XM_017029618.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0X00007791.hg.1 | ENST00000445417.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009897.hg.1 | ENST00000560960.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014878.hg.1 | ENST00000380526.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014878.hg.1 | NR_002830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014878.hg.1 | NR_002830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014878.hg.1 | NR_002830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X011607.hg.1 | TC0X000870.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X011607.hg.1 | ENST00000380523.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X011607.hg.1 | ENST00000398355.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X011607.hg.1 | NM_001042479.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X011607.hg.1 | NM_001042480.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X011607.hg.1 | NM_017856.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008047.hg.1 | ENST00000569855.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008047.hg.1 | ENST00000569855.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000332885.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000332885.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000332885.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000332885.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000332885.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000380523.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000380523.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000380523.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000380523.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000380523.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000380523.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000380523.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000380523.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000380523.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000380523.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000398355.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000398355.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000398355.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000398355.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000398355.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000398355.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000398355.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000460203.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000460203.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000460203.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000477386.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000477386.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000477386.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000477386.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_001042479.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_001042479.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_001042479.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_001042479.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_001042479.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_001042479.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_001042479.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_001042479.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_001042479.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_001042479.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_001042480.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_001042480.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_001042480.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_001042480.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_001042480.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_001042480.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_001042480.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_001042480.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_001042480.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_017856.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_017856.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_017856.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_017856.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_017856.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_017856.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_017856.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_017856.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_017856.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_017856.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_017856.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007791.hg.1 | ENST00000445417.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000332885.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000332885.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000332885.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000332885.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000332885.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000380523.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000380523.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000380523.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000380523.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000380523.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000380523.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000380523.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000380523.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000380523.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000380523.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000398355.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000398355.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000398355.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000398355.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000398355.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000398355.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000398355.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000460203.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000460203.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000460203.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000477386.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000477386.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000477386.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000477386.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_001042479.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_001042479.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_001042479.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_001042479.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_001042479.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_001042479.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_001042479.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_001042479.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_001042479.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_001042479.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_001042480.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_001042480.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_001042480.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_001042480.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_001042480.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_001042480.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_001042480.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_001042480.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_001042480.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_017856.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_017856.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_017856.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_017856.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_017856.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_017856.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_017856.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_017856.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_017856.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_017856.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_017856.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008047.hg.1 | ENST00000569855.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008047.hg.1 | ENST00000569855.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014878.hg.1 | ENST00000380526.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014878.hg.1 | NR_002830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014878.hg.1 | NR_002830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014878.hg.1 | NR_002830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009897.hg.1 | ENST00000560960.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00162416.hg.1 | ENST00000332885.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00162416.hg.1 | ENST00000380523.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00162416.hg.1 | ENST00000398355.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00162416.hg.1 | ENST00000477386.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00162416.hg.1 | NM_001042479.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00162416.hg.1 | NM_001042480.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00162416.hg.1 | NM_017856.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00162416.hg.1 | ENST00000332885.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00162416.hg.1 | ENST00000380523.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00162416.hg.1 | ENST00000398355.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00162416.hg.1 | ENST00000477386.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00162416.hg.1 | NM_001042479.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00162416.hg.1 | NM_001042480.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00162416.hg.1 | NM_017856.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00162416.hg.1 | TC0X00009107.hg.1 | Clariom D TC |
