Assay Details
| Gene Symbol: KRT16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005557.3 | NP_005548.2 | 1 | 53 | 94 | |
| GenBank mRNA | DC394235.1 | - | 1 | 53 | 94 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17001514.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_005557.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
morphogenesis of an epithelium
inflammatory response
cytoskeleton organization
aging
cell proliferation
epidermis development
keratinocyte differentiation
negative regulation of cell migration
keratinization
hair cycle
innate immune response
intermediate filament cytoskeleton organization
keratinocyte migration
establishment of skin barrier
structural constituent of cytoskeleton
protein binding
morphogenesis of an epithelium
inflammatory response
cytoskeleton organization
aging
cell proliferation
epidermis development
keratinocyte differentiation
negative regulation of cell migration
keratinization
hair cycle
innate immune response
intermediate filament cytoskeleton organization
keratinocyte migration
establishment of skin barrier
structural constituent of cytoskeleton
protein binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700010681.hg.1 | ENST00000301653.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010681.hg.1 | ENST00000301653.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010681.hg.1 | ENST00000301653.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010681.hg.1 | ENST00000301653.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010681.hg.1 | ENST00000301653.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010681.hg.1 | ENST00000301653.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010681.hg.1 | ENST00000301653.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010681.hg.1 | ENST00000301653.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010681.hg.1 | ENST00000301653.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010681.hg.1 | ENST00000301653.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010681.hg.1 | ENST00000301653.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010681.hg.1 | ENST00000301653.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010681.hg.1 | ENST00000301653.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010681.hg.1 | ENST00000588319.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010681.hg.1 | ENST00000588319.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010681.hg.1 | ENST00000588319.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010681.hg.1 | ENST00000588319.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010681.hg.1 | ENST00000588319.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010681.hg.1 | ENST00000590990.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010681.hg.1 | ENST00000590990.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010681.hg.1 | ENST00000590990.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010681.hg.1 | ENST00000593067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010681.hg.1 | ENST00000593067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010681.hg.1 | ENST00000593067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010681.hg.1 | ENST00000593067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010681.hg.1 | ENST00000593067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010681.hg.1 | ENST00000593067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010681.hg.1 | ENST00000593067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010681.hg.1 | ENST00000593067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010681.hg.1 | NM_005557.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010681.hg.1 | NM_005557.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010681.hg.1 | NM_005557.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010681.hg.1 | NM_005557.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010681.hg.1 | NM_005557.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010681.hg.1 | NM_005557.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010681.hg.1 | NM_005557.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010681.hg.1 | NM_005557.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010681.hg.1 | NM_005557.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010681.hg.1 | NM_005557.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010681.hg.1 | NM_005557.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010681.hg.1 | NM_005557.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010681.hg.1 | NM_005557.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010681.hg.1 | NM_005557.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17020054.hg.1 | NM_005557.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17020054.hg.1 | TC17001514.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17020051.hg.1 | ENST00000301653.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17020051.hg.1 | NM_005557.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17020051.hg.1 | TC17001514.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1700010681.hg.1 | ENST00000301653.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010681.hg.1 | ENST00000301653.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010681.hg.1 | ENST00000301653.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010681.hg.1 | ENST00000301653.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010681.hg.1 | ENST00000301653.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010681.hg.1 | ENST00000301653.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010681.hg.1 | ENST00000301653.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010681.hg.1 | ENST00000301653.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010681.hg.1 | ENST00000301653.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010681.hg.1 | ENST00000301653.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010681.hg.1 | ENST00000301653.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010681.hg.1 | ENST00000301653.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010681.hg.1 | ENST00000301653.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010681.hg.1 | ENST00000588319.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010681.hg.1 | ENST00000588319.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010681.hg.1 | ENST00000588319.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010681.hg.1 | ENST00000588319.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010681.hg.1 | ENST00000588319.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010681.hg.1 | ENST00000590990.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010681.hg.1 | ENST00000590990.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010681.hg.1 | ENST00000590990.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010681.hg.1 | ENST00000593067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010681.hg.1 | ENST00000593067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010681.hg.1 | ENST00000593067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010681.hg.1 | ENST00000593067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010681.hg.1 | ENST00000593067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010681.hg.1 | ENST00000593067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010681.hg.1 | ENST00000593067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010681.hg.1 | ENST00000593067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010681.hg.1 | NM_005557.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010681.hg.1 | NM_005557.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010681.hg.1 | NM_005557.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010681.hg.1 | NM_005557.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010681.hg.1 | NM_005557.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010681.hg.1 | NM_005557.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010681.hg.1 | NM_005557.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010681.hg.1 | NM_005557.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010681.hg.1 | NM_005557.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010681.hg.1 | NM_005557.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010681.hg.1 | NM_005557.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010681.hg.1 | NM_005557.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010681.hg.1 | NM_005557.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010681.hg.1 | NM_005557.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700184517.hg.1 | ENST00000301653.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184517.hg.1 | ENST00000588319.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184517.hg.1 | ENST00000590990.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184517.hg.1 | NM_005557.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700184517.hg.1 | ENST00000301653.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184517.hg.1 | ENST00000588319.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184517.hg.1 | ENST00000590990.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184517.hg.1 | NM_005557.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700184517.hg.1 | TC1700010681.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700184515.hg.1 | ENST00000301653.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184515.hg.1 | ENST00000588319.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184515.hg.1 | NM_005557.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700184515.hg.1 | ENST00000301653.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184515.hg.1 | ENST00000588319.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184515.hg.1 | NM_005557.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700184515.hg.1 | TC1700010681.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700184516.hg.1 | ENST00000301653.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184516.hg.1 | ENST00000588319.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184516.hg.1 | ENST00000590990.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184516.hg.1 | NM_005557.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700184516.hg.1 | ENST00000301653.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184516.hg.1 | ENST00000588319.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184516.hg.1 | ENST00000590990.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184516.hg.1 | NM_005557.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700184516.hg.1 | TC1700010681.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700184522.hg.1 | ENST00000593067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184522.hg.1 | NM_005557.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700184522.hg.1 | ENST00000593067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184522.hg.1 | NM_005557.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700184522.hg.1 | TC1700010681.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 141639 | - | - |

KRT16
Genomic Map