Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005299.hg.1 |
TC22001466.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005299.hg.1 |
ENST00000359114.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005299.hg.1 |
NM_024313.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005298.hg.1 |
TC22001466.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005298.hg.1 |
ENST00000359114.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005298.hg.1 |
NM_024313.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200151342.hg.1 |
TC2200007313.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200151342.hg.1 |
ENST00000359114.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151342.hg.1 |
ENST00000438329.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151342.hg.1 |
ENST00000455236.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151342.hg.1 |
ENST00000468597.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151342.hg.1 |
ENST00000474032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151342.hg.1 |
ENST00000611699.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151342.hg.1 |
NM_024313.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200151342.hg.1 |
ENST00000359114.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151342.hg.1 |
ENST00000438329.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151342.hg.1 |
ENST00000455236.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151342.hg.1 |
ENST00000468597.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151342.hg.1 |
ENST00000474032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151342.hg.1 |
ENST00000611699.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151342.hg.1 |
NM_024313.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200151340.hg.1 |
TC2200007313.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200151340.hg.1 |
ENST00000359114.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151340.hg.1 |
ENST00000438329.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151340.hg.1 |
ENST00000468597.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151340.hg.1 |
ENST00000474032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151340.hg.1 |
ENST00000611699.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151340.hg.1 |
NM_024313.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200151340.hg.1 |
ENST00000359114.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151340.hg.1 |
ENST00000438329.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151340.hg.1 |
ENST00000468597.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151340.hg.1 |
ENST00000474032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151340.hg.1 |
ENST00000611699.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151340.hg.1 |
NM_024313.2 |
Refseq |